Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201823572-201823831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
UniGene
Hs.348711
Gene Chromosome Location
Chr.2: 201790324-201895550 on Build GRCh38
Transcripts
NM_001261435.1 NM_001261436.1 NM_139158.2 XM_005246781.3 XM_005246782.4 XM_011511650.2 XM_011511652.1 XM_011511654.2 XM_011511655.2 XM_011511656.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7077816 c.556G>A p.A186T Chr.2: 201823677-201823677 on Build GRCh38
6625407 c.406C>T p.Q136* Chr.2: 201823680-201823680 on Build GRCh38
6244897 c.431G>T p.G144V Chr.2: 201823705-201823705 on Build GRCh38
6305736 c.414G>T p.M138I Chr.2: 201823688-201823688 on Build GRCh38
6625408 c.559C>T p.Q187* Chr.2: 201823680-201823680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745755414 Chr.2: 201823704-201823704 on Build GRCh38
rs1050760623 Chr.2: 201823732-201823732 on Build GRCh38
rs768185763 Chr.2: 201823728-201823728 on Build GRCh38
rs777016053 Chr.2: 201823768-201823768 on Build GRCh38
rs542392513 Chr.2: 201823625-201823625 on Build GRCh38
rs765790847 Chr.2: 201823688-201823688 on Build GRCh38
rs191446880 Chr.2: 201823674-201823674 on Build GRCh38
rs201806765 Chr.2: 201823735-201823735 on Build GRCh38
rs778216084 Chr.2: 201823699-201823699 on Build GRCh38
rs748169761 Chr.2: 201823727-201823727 on Build GRCh38
rs369220812 Chr.2: 201823648-201823648 on Build GRCh38
rs774290192 Chr.2: 201823663-201823663 on Build GRCh38
rs869241685 Chr.2: 201823765-201823765 on Build GRCh38
rs766563923 Chr.2: 201823671-201823671 on Build GRCh38
rs780672146 Chr.2: 201823646-201823646 on Build GRCh38
rs937321906 Chr.2: 201823613-201823613 on Build GRCh38
rs753282798 Chr.2: 201823696-201823696 on Build GRCh38
rs571065515 Chr.2: 201823627-201823627 on Build GRCh38
rs371018921 Chr.2: 201823650-201823650 on Build GRCh38
rs114832060 Chr.2: 201823748-201823748 on Build GRCh38
rs377517876 Chr.2: 201823651-201823651 on Build GRCh38
rs771749929 Chr.2: 201823737-201823737 on Build GRCh38
rs147107869 Chr.2: 201823697-201823697 on Build GRCh38
rs869305975 Chr.2: 201823767-201823767 on Build GRCh38
rs890208304 Chr.2: 201823684-201823684 on Build GRCh38
rs199869929 Chr.2: 201823668-201823668 on Build GRCh38
rs551216043 Chr.2: 201823784-201823784 on Build GRCh38
rs759652640 Chr.2: 201823779-201823779 on Build GRCh38
rs780293255 Chr.2: 201823622-201823622 on Build GRCh38
rs761014003 Chr.2: 201823669-201823669 on Build GRCh38
rs373678952 Chr.2: 201823634-201823634 on Build GRCh38
rs373766977 Chr.2: 201823719-201823719 on Build GRCh38
rs767348456 Chr.2: 201823666-201823666 on Build GRCh38
rs761474386 Chr.2: 201823665-201823665 on Build GRCh38
rs755801738 Chr.2: 201823716-201823716 on Build GRCh38
rs766757848 Chr.2: 201823750-201823750 on Build GRCh38
rs935552500 Chr.2: 201823626-201823626 on Build GRCh38
rs745500022 Chr.2: 201823705-201823705 on Build GRCh38
rs755520069 Chr.2: 201823686-201823686 on Build GRCh38
rs374059521 Chr.2: 201823736-201823736 on Build GRCh38
rs530535751 Chr.2: 201823611-201823611 on Build GRCh38
rs1041576494 Chr.2: 201823675-201823675 on Build GRCh38
rs768625842 Chr.2: 201823653-201823653 on Build GRCh38
rs748510409 Chr.2: 201823641-201823641 on Build GRCh38
rs772071625 Chr.2: 201823753-201823753 on Build GRCh38

View Full Product Details