Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 32599971-32600196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD27
Entrez Gene ID
84079
Gene Name
ankyrin repeat domain 27
Gene Aliases
PP12899, VARP
UniGene
Hs.59236
Gene Chromosome Location
Chr.19: 32597001-32675215 on Build GRCh38
Transcripts
NM_032139.2 XM_006723412.2 XM_017027349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4076721 c.2820C>T p.Y940Y Chr.19: 32599998-32599998 on Build GRCh38
7430445 c.2814G>C p.Q938H Chr.19: 32600004-32600004 on Build GRCh38
5071948 c.2768-260_2846+71del410 p.? Chr.19: 32599901-32600310 on Build GRCh38
6242568 c.2846A>G p.K949R Chr.19: 32599972-32599972 on Build GRCh38
439205 c.2790A>G p.L930L Chr.19: 32600028-32600028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774787728 Chr.19: 32600000-32600000 on Build GRCh38
rs747175132 Chr.19: 32600021-32600021 on Build GRCh38
rs753702581 Chr.19: 32600092-32600092 on Build GRCh38
rs141361167 Chr.19: 32600037-32600037 on Build GRCh38
rs766077204 Chr.19: 32600078-32600083 on Build GRCh38
rs762280073 Chr.19: 32600036-32600037 on Build GRCh38
rs766979310 Chr.19: 32600073-32600073 on Build GRCh38
rs777034289 Chr.19: 32600033-32600033 on Build GRCh38
rs1050484626 Chr.19: 32600149-32600149 on Build GRCh38
rs746126809 Chr.19: 32600034-32600034 on Build GRCh38
rs765952352 Chr.19: 32600088-32600088 on Build GRCh38
rs754949637 Chr.19: 32599998-32599998 on Build GRCh38
rs770893802 Chr.19: 32600022-32600022 on Build GRCh38
rs376694174 Chr.19: 32600054-32600054 on Build GRCh38
rs369714895 Chr.19: 32600082-32600082 on Build GRCh38
rs919756875 Chr.19: 32600136-32600136 on Build GRCh38
rs747195938 Chr.19: 32600155-32600155 on Build GRCh38
rs182859468 Chr.19: 32600138-32600138 on Build GRCh38
rs760339123 Chr.19: 32600086-32600086 on Build GRCh38
rs943655609 Chr.19: 32600009-32600009 on Build GRCh38
rs763158972 Chr.19: 32600044-32600044 on Build GRCh38
rs372688508 Chr.19: 32600148-32600148 on Build GRCh38
rs770093544 Chr.19: 32600035-32600035 on Build GRCh38
rs761431174 Chr.19: 32600066-32600066 on Build GRCh38
rs778716421 Chr.19: 32600011-32600011 on Build GRCh38
rs769272187 Chr.19: 32600045-32600045 on Build GRCh38
rs973899814 Chr.19: 32600102-32600102 on Build GRCh38

View Full Product Details