Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58494259-58494532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7447670 c.1033G>A p.V345I Chr.8: 58494512-58494512 on Build GRCh38
3900802 c.1014A>G p.A338A Chr.8: 58494531-58494531 on Build GRCh38
6484327 c.1013C>G p.A338G Chr.8: 58494532-58494532 on Build GRCh38
1489354 c.1038A>C p.L346F Chr.8: 58494507-58494507 on Build GRCh38
6181228 c.1018C>A p.L340M Chr.8: 58494527-58494527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8192875 Chr.8: 58494506-58494506 on Build GRCh38
rs546018615 Chr.8: 58494384-58494384 on Build GRCh38
rs541420341 Chr.8: 58494487-58494487 on Build GRCh38
rs962210762 Chr.8: 58494408-58494408 on Build GRCh38
rs762703827 Chr.8: 58494491-58494491 on Build GRCh38
rs762206399 Chr.8: 58494457-58494457 on Build GRCh38
rs141672922 Chr.8: 58494422-58494422 on Build GRCh38
rs535788376 Chr.8: 58494381-58494381 on Build GRCh38
rs1028989285 Chr.8: 58494299-58494303 on Build GRCh38
rs768646602 Chr.8: 58494289-58494289 on Build GRCh38
rs747365511 Chr.8: 58494463-58494463 on Build GRCh38
rs769016772 Chr.8: 58494471-58494471 on Build GRCh38
rs180695354 Chr.8: 58494495-58494495 on Build GRCh38
rs3747809 Chr.8: 58494425-58494425 on Build GRCh38
rs551090709 Chr.8: 58494291-58494291 on Build GRCh38
rs759259236 Chr.8: 58494503-58494503 on Build GRCh38
rs138571220 Chr.8: 58494380-58494380 on Build GRCh38
rs1057493086 Chr.8: 58494435-58494435 on Build GRCh38
rs777281553 Chr.8: 58494486-58494486 on Build GRCh38
rs756035111 Chr.8: 58494507-58494507 on Build GRCh38
rs556481820 Chr.8: 58494351-58494351 on Build GRCh38
rs372610666 Chr.8: 58494493-58494493 on Build GRCh38
rs368772513 Chr.8: 58494504-58494504 on Build GRCh38
rs937515892 Chr.8: 58494428-58494428 on Build GRCh38
rs77169716 Chr.8: 58494325-58494325 on Build GRCh38
rs570996706 Chr.8: 58494320-58494320 on Build GRCh38
rs769547952 Chr.8: 58494505-58494505 on Build GRCh38
rs895988054 Chr.8: 58494437-58494437 on Build GRCh38
rs1044798063 Chr.8: 58494423-58494423 on Build GRCh38
rs201568509 Chr.8: 58494490-58494490 on Build GRCh38

View Full Product Details