Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 69632663-69632924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf35
Entrez Gene ID
219738
Gene Name
chromosome 10 open reading frame 35
Gene Aliases
-
UniGene
Hs.522992
Gene Chromosome Location
Chr.10: 69630241-69633599 on Build GRCh38
Transcripts
NM_145306.2 XM_005269606.1 XM_005269608.3 XM_011539455.2 XM_017015836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3440009 c.263C>T p.P88L Chr.10: 69632830-69632830 on Build GRCh38
4015436 c.163G>A p.A55T Chr.10: 69632856-69632856 on Build GRCh38
3751915 c.234C>T p.N78N Chr.10: 69632801-69632801 on Build GRCh38
3751914 c.108C>T p.N36N Chr.10: 69632801-69632801 on Build GRCh38
4179986 c.302C>T p.A101V Chr.10: 69632869-69632869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531876328 Chr.10: 69632767-69632767 on Build GRCh38
rs1026803860 Chr.10: 69632718-69632718 on Build GRCh38
rs762661683 Chr.10: 69632829-69632829 on Build GRCh38
rs748091788 Chr.10: 69632887-69632887 on Build GRCh38
rs767858328 Chr.10: 69632762-69632762 on Build GRCh38
rs761088839 Chr.10: 69632849-69632849 on Build GRCh38
rs772941500 Chr.10: 69632747-69632747 on Build GRCh38
rs757448881 Chr.10: 69632871-69632871 on Build GRCh38
rs773064297 Chr.10: 69632818-69632818 on Build GRCh38
rs764439740 Chr.10: 69632854-69632854 on Build GRCh38
rs201893020 Chr.10: 69632817-69632817 on Build GRCh38
rs768661090 Chr.10: 69632806-69632806 on Build GRCh38
rs41306516 Chr.10: 69632727-69632727 on Build GRCh38
rs12020 Chr.10: 69632801-69632801 on Build GRCh38
rs199529674 Chr.10: 69632808-69632808 on Build GRCh38
rs773930460 Chr.10: 69632838-69632838 on Build GRCh38
rs778953654 Chr.10: 69632876-69632876 on Build GRCh38
rs762673754 Chr.10: 69632851-69632851 on Build GRCh38
rs924954484 Chr.10: 69632853-69632853 on Build GRCh38
rs775308790 Chr.10: 69632792-69632794 on Build GRCh38
rs150058848 Chr.10: 69632730-69632730 on Build GRCh38
rs879061782 Chr.10: 69632695-69632695 on Build GRCh38
rs765980457 Chr.10: 69632755-69632755 on Build GRCh38
rs779021457 Chr.10: 69632800-69632800 on Build GRCh38
rs768377454 Chr.10: 69632743-69632743 on Build GRCh38
rs758351510 Chr.10: 69632881-69632881 on Build GRCh38
rs770644575 Chr.10: 69632906-69632906 on Build GRCh38
rs564809612 Chr.10: 69632742-69632742 on Build GRCh38
rs913640343 Chr.10: 69632821-69632821 on Build GRCh38
rs370741437 Chr.10: 69632898-69632898 on Build GRCh38
rs768706315 Chr.10: 69632731-69632731 on Build GRCh38
rs762608663 Chr.10: 69632754-69632754 on Build GRCh38
rs757397371 Chr.10: 69632794-69632794 on Build GRCh38
rs779915874 Chr.10: 69632741-69632741 on Build GRCh38
rs1051256836 Chr.10: 69632896-69632896 on Build GRCh38
rs144236141 Chr.10: 69632890-69632890 on Build GRCh38
rs760063248 Chr.10: 69632761-69632761 on Build GRCh38
rs923550346 Chr.10: 69632769-69632769 on Build GRCh38
rs752043730 Chr.10: 69632757-69632757 on Build GRCh38
rs376428994 Chr.10: 69632764-69632764 on Build GRCh38
rs370959938 Chr.10: 69632782-69632782 on Build GRCh38
rs770392905 Chr.10: 69632830-69632830 on Build GRCh38
rs201072929 Chr.10: 69632839-69632839 on Build GRCh38
rs769697064 Chr.10: 69632814-69632814 on Build GRCh38
rs769944524 Chr.10: 69632766-69632769 on Build GRCh38
rs2616089 Chr.10: 69632739-69632739 on Build GRCh38
rs934002622 Chr.10: 69632822-69632822 on Build GRCh38
rs781247364 Chr.10: 69632886-69632886 on Build GRCh38
rs138995196 Chr.10: 69632897-69632897 on Build GRCh38
rs878989803 Chr.10: 69632816-69632816 on Build GRCh38
rs2616088 Chr.10: 69632745-69632745 on Build GRCh38
rs750521333 Chr.10: 69632880-69632880 on Build GRCh38
rs777987761 Chr.10: 69632775-69632775 on Build GRCh38
rs753148471 Chr.10: 69632845-69632845 on Build GRCh38
rs1013443029 Chr.10: 69632704-69632704 on Build GRCh38
rs747969242 Chr.10: 69632812-69632812 on Build GRCh38
rs776464745 Chr.10: 69632744-69632744 on Build GRCh38

View Full Product Details