Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35825518-35825775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221B
Entrez Gene ID
392307
Gene Name
family with sequence similarity 221 member B
Gene Aliases
C9orf128
UniGene
Hs.128075
Gene Chromosome Location
Chr.9: 35817017-35829148 on Build GRCh38
Transcripts
NM_001012446.3 NR_052026.1 XM_006716768.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5063965 c.598+2C>T p.? Chr.9: 35825562-35825562 on Build GRCh38
6871195 c.473C>A p.S158* Chr.9: 35825689-35825689 on Build GRCh38
1108842 c.519G>T p.E173D Chr.9: 35825643-35825643 on Build GRCh38
3657269 c.544G>A p.G182R Chr.9: 35825618-35825618 on Build GRCh38
3657277 c.490G>A p.V164I Chr.9: 35825672-35825672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537497131 Chr.9: 35825698-35825698 on Build GRCh38
rs1019701017 Chr.9: 35825739-35825739 on Build GRCh38
rs201802596 Chr.9: 35825545-35825547 on Build GRCh38
rs756455271 Chr.9: 35825602-35825602 on Build GRCh38
rs375534080 Chr.9: 35825612-35825612 on Build GRCh38
rs981788234 Chr.9: 35825592-35825592 on Build GRCh38
rs931862343 Chr.9: 35825665-35825665 on Build GRCh38
rs115407050 Chr.9: 35825730-35825730 on Build GRCh38
rs753055882 Chr.9: 35825600-35825600 on Build GRCh38
rs752927511 Chr.9: 35825724-35825724 on Build GRCh38
rs751671992 Chr.9: 35825578-35825578 on Build GRCh38
rs907721681 Chr.9: 35825599-35825599 on Build GRCh38
rs754306916 Chr.9: 35825610-35825610 on Build GRCh38
rs767820159 Chr.9: 35825595-35825595 on Build GRCh38
rs566673349 Chr.9: 35825550-35825550 on Build GRCh38
rs201425299 Chr.9: 35825709-35825709 on Build GRCh38
rs779597219 Chr.9: 35825614-35825614 on Build GRCh38
rs766706252 Chr.9: 35825636-35825636 on Build GRCh38
rs374974977 Chr.9: 35825630-35825630 on Build GRCh38
rs764461401 Chr.9: 35825748-35825748 on Build GRCh38
rs759900623 Chr.9: 35825588-35825588 on Build GRCh38
rs79210384 Chr.9: 35825654-35825654 on Build GRCh38
rs905793541 Chr.9: 35825579-35825579 on Build GRCh38
rs202228927 Chr.9: 35825664-35825664 on Build GRCh38
rs777498192 Chr.9: 35825544-35825545 on Build GRCh38
rs762943958 Chr.9: 35825553-35825553 on Build GRCh38
rs746490281 Chr.9: 35825623-35825623 on Build GRCh38
rs373107675 Chr.9: 35825655-35825655 on Build GRCh38
rs763002885 Chr.9: 35825672-35825672 on Build GRCh38
rs1013714518 Chr.9: 35825659-35825659 on Build GRCh38
rs773159548 Chr.9: 35825552-35825552 on Build GRCh38
rs940521288 Chr.9: 35825607-35825607 on Build GRCh38
rs774616330 Chr.9: 35825699-35825699 on Build GRCh38
rs766506704 Chr.9: 35825565-35825565 on Build GRCh38
rs770984765 Chr.9: 35825676-35825676 on Build GRCh38
rs778177155 Chr.9: 35825605-35825605 on Build GRCh38
rs538950774 Chr.9: 35825583-35825583 on Build GRCh38
rs780775095 Chr.9: 35825645-35825645 on Build GRCh38
rs759668983 Chr.9: 35825703-35825703 on Build GRCh38
rs1049770753 Chr.9: 35825707-35825707 on Build GRCh38

View Full Product Details