Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27879533-27879758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
UniGene
Hs.10649
Gene Chromosome Location
Chr.1: 27872531-27886685 on Build GRCh38
Transcripts
NM_001039477.2 NM_001105556.2 NM_001286113.1 NM_001286115.1 NM_004848.3 XM_005246041.3 XM_006711050.1 XM_011542445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5516149 c.260_261delCA p.Q88fs*3 Chr.1: 27879668-27879669 on Build GRCh38
4482358 c.259C>A p.P87T Chr.1: 27879667-27879667 on Build GRCh38
7353144 c.331T>C p.F111L Chr.1: 27879739-27879739 on Build GRCh38
77376 c.302C>T p.T101I Chr.1: 27879710-27879710 on Build GRCh38
4895883 c.279G>A p.L93L Chr.1: 27879687-27879687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768112330 Chr.1: 27879688-27879688 on Build GRCh38
rs763974146 Chr.1: 27879653-27879653 on Build GRCh38
rs1024366055 Chr.1: 27879559-27879559 on Build GRCh38
rs146409624 Chr.1: 27879658-27879658 on Build GRCh38
rs199929444 Chr.1: 27879695-27879696 on Build GRCh38
rs985951269 Chr.1: 27879668-27879668 on Build GRCh38
rs763173212 Chr.1: 27879731-27879731 on Build GRCh38
rs143326302 Chr.1: 27879706-27879706 on Build GRCh38
rs372068799 Chr.1: 27879617-27879617 on Build GRCh38
rs895406030 Chr.1: 27879734-27879734 on Build GRCh38
rs145503231 Chr.1: 27879697-27879697 on Build GRCh38
rs1015985965 Chr.1: 27879588-27879588 on Build GRCh38
rs1056608063 Chr.1: 27879717-27879717 on Build GRCh38
rs766571282 Chr.1: 27879607-27879607 on Build GRCh38
rs753781345 Chr.1: 27879640-27879640 on Build GRCh38
rs1004181083 Chr.1: 27879580-27879580 on Build GRCh38
rs186451576 Chr.1: 27879606-27879606 on Build GRCh38
rs779512916 Chr.1: 27879665-27879665 on Build GRCh38
rs750799391 Chr.1: 27879599-27879599 on Build GRCh38
rs573092977 Chr.1: 27879577-27879577 on Build GRCh38
rs879034401 Chr.1: 27879645-27879645 on Build GRCh38
rs778739366 Chr.1: 27879657-27879657 on Build GRCh38
rs757308881 Chr.1: 27879649-27879649 on Build GRCh38
rs35953961 Chr.1: 27879587-27879587 on Build GRCh38
rs776137764 Chr.1: 27879696-27879696 on Build GRCh38
rs944417245 Chr.1: 27879691-27879691 on Build GRCh38
rs375175632 Chr.1: 27879623-27879623 on Build GRCh38
rs746675739 Chr.1: 27879675-27879675 on Build GRCh38
rs151326930 Chr.1: 27879597-27879597 on Build GRCh38
rs770357358 Chr.1: 27879699-27879699 on Build GRCh38
rs771740449 Chr.1: 27879663-27879663 on Build GRCh38
rs971440934 Chr.1: 27879565-27879565 on Build GRCh38
rs920520443 Chr.1: 27879625-27879625 on Build GRCh38
rs987424379 Chr.1: 27879560-27879560 on Build GRCh38
rs781454382 Chr.1: 27879639-27879639 on Build GRCh38

View Full Product Details