Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232543574-232543831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNG
Entrez Gene ID
1146
Gene Name
cholinergic receptor nicotinic gamma subunit
Gene Aliases
ACHRG
UniGene
Hs.248101
Gene Chromosome Location
Chr.2: 232539727-232546328 on Build GRCh38
Transcripts
NM_005199.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5549573 c.1019C>A p.A340D Chr.2: 232543683-232543683 on Build GRCh38
4611795 c.1000_1001insCA p.S338fs*12 Chr.2: 232543664-232543665 on Build GRCh38
5896605 c.1025G>A p.G342E Chr.2: 232543689-232543689 on Build GRCh38
1669850 c.994C>T p.R332W Chr.2: 232543658-232543658 on Build GRCh38
1406243 c.1030C>T p.R344C Chr.2: 232543694-232543694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752487562 Chr.2: 232543685-232543685 on Build GRCh38
rs571741639 Chr.2: 232543701-232543701 on Build GRCh38
rs746613858 Chr.2: 232543719-232543719 on Build GRCh38
rs367569763 Chr.2: 232543740-232543740 on Build GRCh38
rs866314593 Chr.2: 232543812-232543812 on Build GRCh38
rs772412938 Chr.2: 232543711-232543711 on Build GRCh38
rs997856549 Chr.2: 232543716-232543716 on Build GRCh38
rs779291270 Chr.2: 232543749-232543749 on Build GRCh38
rs758713523 Chr.2: 232543750-232543750 on Build GRCh38
rs577754250 Chr.2: 232543732-232543732 on Build GRCh38
rs769092268 Chr.2: 232543643-232543643 on Build GRCh38
rs551573361 Chr.2: 232543773-232543773 on Build GRCh38
rs542961885 Chr.2: 232543759-232543759 on Build GRCh38
rs540701045 Chr.2: 232543720-232543720 on Build GRCh38
rs756205168 Chr.2: 232543693-232543693 on Build GRCh38
rs528525264 Chr.2: 232543769-232543769 on Build GRCh38
rs138637669 Chr.2: 232543625-232543625 on Build GRCh38
rs57945834 Chr.2: 232543763-232543763 on Build GRCh38
rs72991926 Chr.2: 232543804-232543804 on Build GRCh38
rs370687934 Chr.2: 232543709-232543709 on Build GRCh38
rs753880381 Chr.2: 232543695-232543695 on Build GRCh38
rs772992979 Chr.2: 232543655-232543655 on Build GRCh38
rs779181232 Chr.2: 232543645-232543645 on Build GRCh38
rs772316123 Chr.2: 232543683-232543683 on Build GRCh38
rs771587350 Chr.2: 232543760-232543760 on Build GRCh38
rs140175279 Chr.2: 232543739-232543739 on Build GRCh38
rs771680359 Chr.2: 232543654-232543654 on Build GRCh38
rs778440648 Chr.2: 232543605-232543605 on Build GRCh38
rs75369104 Chr.2: 232543615-232543615 on Build GRCh38
rs771407292 Chr.2: 232543647-232543648 on Build GRCh38
rs149535359 Chr.2: 232543624-232543624 on Build GRCh38
rs777583959 Chr.2: 232543718-232543718 on Build GRCh38
rs141485470 Chr.2: 232543686-232543686 on Build GRCh38
rs113978875 Chr.2: 232543660-232543660 on Build GRCh38
rs753762944 Chr.2: 232543601-232543601 on Build GRCh38
rs982792051 Chr.2: 232543768-232543768 on Build GRCh38
rs769976309 Chr.2: 232543805-232543805 on Build GRCh38
rs372687247 Chr.2: 232543647-232543647 on Build GRCh38
rs746007534 Chr.2: 232543626-232543626 on Build GRCh38
rs767099064 Chr.2: 232543633-232543633 on Build GRCh38
rs375987895 Chr.2: 232543659-232543659 on Build GRCh38
rs554907332 Chr.2: 232543694-232543694 on Build GRCh38
rs776322041 Chr.2: 232543721-232543721 on Build GRCh38
rs73995686 Chr.2: 232543748-232543748 on Build GRCh38
rs934289837 Chr.2: 232543772-232543772 on Build GRCh38
rs376907664 Chr.2: 232543730-232543730 on Build GRCh38
rs144251231 Chr.2: 232543724-232543724 on Build GRCh38
rs750582540 Chr.2: 232543688-232543688 on Build GRCh38
rs764892395 Chr.2: 232543620-232543620 on Build GRCh38
rs200922542 Chr.2: 232543734-232543734 on Build GRCh38
rs756184613 Chr.2: 232543631-232543631 on Build GRCh38
rs750673627 Chr.2: 232543735-232543735 on Build GRCh38
rs567899708 Chr.2: 232543658-232543658 on Build GRCh38
rs970988519 Chr.2: 232543764-232543764 on Build GRCh38
rs776733115 Chr.2: 232543676-232543676 on Build GRCh38
rs775496585 Chr.2: 232543731-232543731 on Build GRCh38
rs1015515236 Chr.2: 232543692-232543692 on Build GRCh38
rs766562368 Chr.2: 232543742-232543742 on Build GRCh38
rs759353019 Chr.2: 232543679-232543679 on Build GRCh38
rs749744672 Chr.2: 232543642-232543642 on Build GRCh38

View Full Product Details