Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 26483097-26483342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCKAR
Entrez Gene ID
886
Gene Name
cholecystokinin A receptor
Gene Aliases
CCK-A, CCK1-R, CCK1R, CCKRA
UniGene
Hs.129
Gene Chromosome Location
Chr.4: 26481396-26490420 on Build GRCh38
Transcripts
NM_000730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
95585 c.712C>T p.Q238* Chr.4: 26483198-26483198 on Build GRCh38
1200122 c.698C>A p.S233Y Chr.4: 26483212-26483212 on Build GRCh38
5885379 c.661C>T p.P221S Chr.4: 26483249-26483249 on Build GRCh38
3133671 c.678G>C p.M226I Chr.4: 26483232-26483232 on Build GRCh38
5496104 c.689G>T p.G230V Chr.4: 26483221-26483221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200826139 Chr.4: 26483216-26483216 on Build GRCh38
rs200794266 Chr.4: 26483237-26483237 on Build GRCh38
rs773043148 Chr.4: 26483259-26483259 on Build GRCh38
rs1040865701 Chr.4: 26483204-26483204 on Build GRCh38
rs745998238 Chr.4: 26483198-26483198 on Build GRCh38
rs748487004 Chr.4: 26483126-26483126 on Build GRCh38
rs752324124 Chr.4: 26483285-26483285 on Build GRCh38
rs528155442 Chr.4: 26483157-26483157 on Build GRCh38
rs766373873 Chr.4: 26483269-26483269 on Build GRCh38
rs200173940 Chr.4: 26483133-26483133 on Build GRCh38
rs554214143 Chr.4: 26483166-26483168 on Build GRCh38
rs778384208 Chr.4: 26483149-26483149 on Build GRCh38
rs201328804 Chr.4: 26483147-26483147 on Build GRCh38
rs201996688 Chr.4: 26483249-26483249 on Build GRCh38
rs140481104 Chr.4: 26483224-26483224 on Build GRCh38
rs747408306 Chr.4: 26483299-26483300 on Build GRCh38
rs1020893615 Chr.4: 26483273-26483273 on Build GRCh38
rs376612950 Chr.4: 26483289-26483289 on Build GRCh38
rs778372953 Chr.4: 26483228-26483230 on Build GRCh38
rs370015751 Chr.4: 26483278-26483278 on Build GRCh38
rs865910758 Chr.4: 26483207-26483207 on Build GRCh38
rs201308743 Chr.4: 26483183-26483183 on Build GRCh38
rs775585316 Chr.4: 26483208-26483208 on Build GRCh38
rs199686239 Chr.4: 26483205-26483205 on Build GRCh38
rs745901045 Chr.4: 26483314-26483314 on Build GRCh38
rs531262435 Chr.4: 26483311-26483311 on Build GRCh38
rs764896911 Chr.4: 26483284-26483284 on Build GRCh38
rs761617002 Chr.4: 26483225-26483225 on Build GRCh38
rs199619228 Chr.4: 26483230-26483230 on Build GRCh38
rs778257879 Chr.4: 26483291-26483291 on Build GRCh38
rs41267457 Chr.4: 26483282-26483282 on Build GRCh38
rs760596555 Chr.4: 26483261-26483261 on Build GRCh38
rs771612341 Chr.4: 26483159-26483159 on Build GRCh38
rs76473139 Chr.4: 26483294-26483294 on Build GRCh38
rs201374430 Chr.4: 26483226-26483226 on Build GRCh38
rs781551824 Chr.4: 26483312-26483312 on Build GRCh38
rs755369240 Chr.4: 26483301-26483302 on Build GRCh38
rs372682450 Chr.4: 26483288-26483288 on Build GRCh38

View Full Product Details