Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 121669851-121670112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA5
Entrez Gene ID
308
Gene Name
annexin A5
Gene Aliases
ANX5, ENX2, HEL-S-7, PP4, RPRGL3
UniGene
Hs.480653
Gene Chromosome Location
Chr.4: 121667997-121696992 on Build GRCh38
Transcripts
NM_001154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601323 c.734G>A p.R245Q Chr.4: 121670000-121670000 on Build GRCh38
7121470 c.763C>A p.L255I Chr.4: 121669971-121669971 on Build GRCh38
6485930 c.749A>T p.Y250F Chr.4: 121669985-121669985 on Build GRCh38
732802 c.750C>T p.Y250Y Chr.4: 121669984-121669984 on Build GRCh38
732804 c.751C>T p.L251F Chr.4: 121669983-121669983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950591234 Chr.4: 121669997-121669997 on Build GRCh38
rs778014776 Chr.4: 121669925-121669928 on Build GRCh38
rs770946632 Chr.4: 121669982-121669989 on Build GRCh38
rs748629904 Chr.4: 121669971-121669971 on Build GRCh38
rs199857216 Chr.4: 121670053-121670053 on Build GRCh38
rs761682710 Chr.4: 121669924-121669924 on Build GRCh38
rs955404654 Chr.4: 121670070-121670070 on Build GRCh38
rs373965710 Chr.4: 121670001-121670001 on Build GRCh38
rs753771645 Chr.4: 121669916-121669916 on Build GRCh38
rs531169706 Chr.4: 121670024-121670026 on Build GRCh38
rs765028408 Chr.4: 121669929-121669929 on Build GRCh38
rs140435209 Chr.4: 121669909-121669909 on Build GRCh38
rs768166486 Chr.4: 121670052-121670052 on Build GRCh38
rs750217568 Chr.4: 121670058-121670058 on Build GRCh38
rs746183924 Chr.4: 121669988-121669988 on Build GRCh38
rs878912992 Chr.4: 121669950-121669950 on Build GRCh38
rs1007602770 Chr.4: 121669898-121669898 on Build GRCh38
rs751655674 Chr.4: 121669940-121669940 on Build GRCh38
rs369345487 Chr.4: 121669930-121669930 on Build GRCh38
rs1018101783 Chr.4: 121669914-121669914 on Build GRCh38
rs765142086 Chr.4: 121670057-121670057 on Build GRCh38
rs371378787 Chr.4: 121670026-121670026 on Build GRCh38
rs376983993 Chr.4: 121669935-121669935 on Build GRCh38
rs976069228 Chr.4: 121669956-121669956 on Build GRCh38
rs372823972 Chr.4: 121669931-121669931 on Build GRCh38
rs775164812 Chr.4: 121670048-121670048 on Build GRCh38
rs780469318 Chr.4: 121669939-121669939 on Build GRCh38
rs34303326 Chr.4: 121669908-121669909 on Build GRCh38
rs145513784 Chr.4: 121670000-121670000 on Build GRCh38
rs749339767 Chr.4: 121669949-121669953 on Build GRCh38
rs150126476 Chr.4: 121669963-121669963 on Build GRCh38
rs111418524 Chr.4: 121669998-121669998 on Build GRCh38
rs565568299 Chr.4: 121670049-121670049 on Build GRCh38
rs1029471397 Chr.4: 121670055-121670055 on Build GRCh38
rs771692257 Chr.4: 121670028-121670028 on Build GRCh38
rs368879357 Chr.4: 121669959-121669959 on Build GRCh38
rs761747760 Chr.4: 121670056-121670056 on Build GRCh38

View Full Product Details