Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 239908890-239909129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM3
Entrez Gene ID
1131
Gene Name
cholinergic receptor muscarinic 3
Gene Aliases
EGBRS, HM3, PBS
UniGene
Hs.155736
Gene Chromosome Location
Chr.1: 239386565-239915450 on Build GRCh38
Transcripts
NM_000740.2 XM_005273032.3 XM_011544041.2 XM_011544043.2 XM_011544044.2 XM_011544045.2 XM_011544046.2 XM_011544047.2 XM_017000152.1 XM_017000153.1 XM_017000154.1 XM_017000155.1 XM_017000156.1 XM_017000157.1 XM_017000158.1 XM_017000159.1 XM_017000160.1 XM_017000161.1 XM_017000162.1 XM_017000163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3689469 c.1466C>T p.A489V Chr.1: 239908917-239908917 on Build GRCh38
6035031 c.1574T>A p.F525Y Chr.1: 239909025-239909025 on Build GRCh38
4165098 c.1579A>T p.N527Y Chr.1: 239909030-239909030 on Build GRCh38
6056095 c.1443G>T p.M481I Chr.1: 239908894-239908894 on Build GRCh38
4914941 c.1593G>C p.W531C Chr.1: 239909044-239909044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755937525 Chr.1: 239909001-239909001 on Build GRCh38
rs201554378 Chr.1: 239909011-239909011 on Build GRCh38
rs200598479 Chr.1: 239909109-239909109 on Build GRCh38
rs760985673 Chr.1: 239909065-239909065 on Build GRCh38
rs765946218 Chr.1: 239909077-239909077 on Build GRCh38
rs745877258 Chr.1: 239909048-239909048 on Build GRCh38
rs201983181 Chr.1: 239909022-239909022 on Build GRCh38
rs144917572 Chr.1: 239908938-239908938 on Build GRCh38
rs779167184 Chr.1: 239909023-239909023 on Build GRCh38
rs762623405 Chr.1: 239909075-239909075 on Build GRCh38
rs752495488 Chr.1: 239908975-239908975 on Build GRCh38
rs200534271 Chr.1: 239908983-239908983 on Build GRCh38
rs762495692 Chr.1: 239908918-239908918 on Build GRCh38
rs772511456 Chr.1: 239909054-239909054 on Build GRCh38
rs759076174 Chr.1: 239909087-239909087 on Build GRCh38
rs749499292 Chr.1: 239909021-239909021 on Build GRCh38
rs143305433 Chr.1: 239909089-239909089 on Build GRCh38
rs751027754 Chr.1: 239908936-239908936 on Build GRCh38
rs202061144 Chr.1: 239908960-239908960 on Build GRCh38
rs200289455 Chr.1: 239909067-239909067 on Build GRCh38
rs202208046 Chr.1: 239908934-239908934 on Build GRCh38
rs776058750 Chr.1: 239909059-239909059 on Build GRCh38
rs201424940 Chr.1: 239909071-239909071 on Build GRCh38
rs868683805 Chr.1: 239908966-239908966 on Build GRCh38
rs968297663 Chr.1: 239909056-239909056 on Build GRCh38
rs776927224 Chr.1: 239909074-239909074 on Build GRCh38
rs966153717 Chr.1: 239908921-239908921 on Build GRCh38
rs751087070 Chr.1: 239909083-239909083 on Build GRCh38
rs781128679 Chr.1: 239908944-239908944 on Build GRCh38
rs903730617 Chr.1: 239908935-239908935 on Build GRCh38

View Full Product Details