Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35720489-35720736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL5
Entrez Gene ID
80831
Gene Name
apolipoprotein L5
Gene Aliases
APOL-V, APOLV
UniGene
Hs.326561
Gene Chromosome Location
Chr.22: 35708424-35730279 on Build GRCh38
Transcripts
NM_030642.1 XM_006724321.3 XM_017028945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926137 c.81G>T p.M27I Chr.22: 35720593-35720593 on Build GRCh38
3149323 c.88C>T p.R30* Chr.22: 35720600-35720600 on Build GRCh38
444927 c.76G>A p.E26K Chr.22: 35720588-35720588 on Build GRCh38
1327249 c.112G>A p.V38I Chr.22: 35720624-35720624 on Build GRCh38
1033774 c.68G>T p.G23V Chr.22: 35720580-35720580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025103766 Chr.22: 35720659-35720659 on Build GRCh38
rs567801962 Chr.22: 35720688-35720688 on Build GRCh38
rs779810376 Chr.22: 35720597-35720597 on Build GRCh38
rs757956930 Chr.22: 35720653-35720653 on Build GRCh38
rs370168171 Chr.22: 35720673-35720673 on Build GRCh38
rs568136674 Chr.22: 35720600-35720600 on Build GRCh38
rs757136137 Chr.22: 35720567-35720567 on Build GRCh38
rs375249154 Chr.22: 35720530-35720530 on Build GRCh38
rs915773013 Chr.22: 35720521-35720521 on Build GRCh38
rs149294583 Chr.22: 35720647-35720647 on Build GRCh38
rs765397112 Chr.22: 35720545-35720545 on Build GRCh38
rs368877696 Chr.22: 35720666-35720666 on Build GRCh38
rs749176300 Chr.22: 35720672-35720672 on Build GRCh38
rs984518106 Chr.22: 35720519-35720519 on Build GRCh38
rs182057020 Chr.22: 35720655-35720655 on Build GRCh38
rs763071253 Chr.22: 35720634-35720634 on Build GRCh38
rs376463998 Chr.22: 35720573-35720573 on Build GRCh38
rs757403130 Chr.22: 35720644-35720644 on Build GRCh38
rs550141392 Chr.22: 35720517-35720517 on Build GRCh38
rs768079193 Chr.22: 35720531-35720531 on Build GRCh38
rs569886157 Chr.22: 35720574-35720574 on Build GRCh38
rs772650900 Chr.22: 35720557-35720557 on Build GRCh38
rs892435768 Chr.22: 35720548-35720548 on Build GRCh38
rs889311918 Chr.22: 35720612-35720612 on Build GRCh38
rs181607198 Chr.22: 35720518-35720518 on Build GRCh38
rs754559059 Chr.22: 35720679-35720679 on Build GRCh38
rs777145664 Chr.22: 35720628-35720628 on Build GRCh38
rs373464617 Chr.22: 35720523-35720523 on Build GRCh38
rs771644523 Chr.22: 35720687-35720687 on Build GRCh38
rs771320900 Chr.22: 35720626-35720626 on Build GRCh38
rs759856437 Chr.22: 35720629-35720629 on Build GRCh38
rs374963485 Chr.22: 35720685-35720685 on Build GRCh38
rs981392827 Chr.22: 35720537-35720537 on Build GRCh38
rs768415716 Chr.22: 35720601-35720601 on Build GRCh38
rs565323452 Chr.22: 35720630-35720630 on Build GRCh38
rs369749894 Chr.22: 35720632-35720632 on Build GRCh38
rs779984317 Chr.22: 35720657-35720657 on Build GRCh38
rs751718007 Chr.22: 35720562-35720562 on Build GRCh38
rs139593449 Chr.22: 35720615-35720615 on Build GRCh38
rs759942591 Chr.22: 35720544-35720544 on Build GRCh38
rs368682678 Chr.22: 35720572-35720572 on Build GRCh38
rs769730762 Chr.22: 35720588-35720588 on Build GRCh38
rs201587067 Chr.22: 35720559-35720559 on Build GRCh38
rs529889714 Chr.22: 35720534-35720534 on Build GRCh38
rs147206206 Chr.22: 35720614-35720614 on Build GRCh38
rs777057908 Chr.22: 35720693-35720693 on Build GRCh38
rs369787137 Chr.22: 35720541-35720541 on Build GRCh38
rs761075874 Chr.22: 35720539-35720539 on Build GRCh38
rs921104028 Chr.22: 35720542-35720542 on Build GRCh38
rs76678345 Chr.22: 35720546-35720546 on Build GRCh38
rs993736490 Chr.22: 35720645-35720645 on Build GRCh38
rs750413929 Chr.22: 35720654-35720654 on Build GRCh38
rs773858698 Chr.22: 35720684-35720684 on Build GRCh38

View Full Product Details