Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 93937645-93937904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB2
Entrez Gene ID
51676
Gene Name
ankyrin repeat and SOCS box containing 2
Gene Aliases
ASB-2
UniGene
Hs.510327
Gene Chromosome Location
Chr.14: 93934153-93976796 on Build GRCh38
Transcripts
NM_001202429.1 NM_016150.4 XM_005267758.3 XM_011536834.2 XM_011536835.2 XM_017021369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6569244 c.1575G>A p.L525L Chr.14: 93937750-93937750 on Build GRCh38
3499223 c.1527C>T p.I509I Chr.14: 93937798-93937798 on Build GRCh38
2252923 c.1538T>C p.L513P Chr.14: 93937787-93937787 on Build GRCh38
3956316 c.1553A>T p.N518I Chr.14: 93937772-93937772 on Build GRCh38
5440334 c.1533C>A p.V511V Chr.14: 93937792-93937792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778220167 Chr.14: 93937820-93937820 on Build GRCh38
rs776097016 Chr.14: 93937868-93937868 on Build GRCh38
rs757402918 Chr.14: 93937880-93937880 on Build GRCh38
rs377415399 Chr.14: 93937810-93937810 on Build GRCh38
rs775174511 Chr.14: 93937738-93937738 on Build GRCh38
rs777896257 Chr.14: 93937772-93937772 on Build GRCh38
rs749193769 Chr.14: 93937854-93937854 on Build GRCh38
rs996402210 Chr.14: 93937774-93937774 on Build GRCh38
rs768776624 Chr.14: 93937784-93937784 on Build GRCh38
rs769737457 Chr.14: 93937713-93937713 on Build GRCh38
rs751671391 Chr.14: 93937828-93937828 on Build GRCh38
rs766791443 Chr.14: 93937686-93937687 on Build GRCh38
rs761556357 Chr.14: 93937794-93937794 on Build GRCh38
rs758623192 Chr.14: 93937771-93937771 on Build GRCh38
rs377328485 Chr.14: 93937690-93937690 on Build GRCh38
rs757501542 Chr.14: 93937831-93937831 on Build GRCh38
rs749167415 Chr.14: 93937782-93937782 on Build GRCh38
rs547067875 Chr.14: 93937840-93937840 on Build GRCh38
rs200072486 Chr.14: 93937717-93937717 on Build GRCh38
rs148796321 Chr.14: 93937817-93937817 on Build GRCh38
rs144514108 Chr.14: 93937779-93937779 on Build GRCh38
rs770864987 Chr.14: 93937773-93937773 on Build GRCh38
rs553513775 Chr.14: 93937800-93937800 on Build GRCh38
rs34365480 Chr.14: 93937871-93937871 on Build GRCh38
rs773065479 Chr.14: 93937842-93937842 on Build GRCh38
rs377227565 Chr.14: 93937877-93937877 on Build GRCh38
rs758649817 Chr.14: 93937818-93937818 on Build GRCh38
rs533852407 Chr.14: 93937763-93937763 on Build GRCh38
rs146138244 Chr.14: 93937798-93937798 on Build GRCh38
rs1037692396 Chr.14: 93937758-93937758 on Build GRCh38
rs772250640 Chr.14: 93937677-93937677 on Build GRCh38
rs764756221 Chr.14: 93937869-93937869 on Build GRCh38
rs779245846 Chr.14: 93937764-93937764 on Build GRCh38
rs779264333 Chr.14: 93937670-93937670 on Build GRCh38
rs933994045 Chr.14: 93937795-93937795 on Build GRCh38
rs763920885 Chr.14: 93937731-93937731 on Build GRCh38
rs530975948 Chr.14: 93937755-93937755 on Build GRCh38
rs773327678 Chr.14: 93937687-93937687 on Build GRCh38
rs770627637 Chr.14: 93937864-93937864 on Build GRCh38
rs753094432 Chr.14: 93937870-93937870 on Build GRCh38
rs113704212 Chr.14: 93937716-93937716 on Build GRCh38
rs201903938 Chr.14: 93937757-93937757 on Build GRCh38
rs567814586 Chr.14: 93937797-93937797 on Build GRCh38
rs774270676 Chr.14: 93937787-93937787 on Build GRCh38
rs767128073 Chr.14: 93937753-93937753 on Build GRCh38
rs764296035 Chr.14: 93937876-93937876 on Build GRCh38
rs139476701 Chr.14: 93937756-93937756 on Build GRCh38
rs748303418 Chr.14: 93937676-93937676 on Build GRCh38
rs765742509 Chr.14: 93937804-93937804 on Build GRCh38
rs886563457 Chr.14: 93937720-93937720 on Build GRCh38
rs781413419 Chr.14: 93937833-93937833 on Build GRCh38
rs748570847 Chr.14: 93937770-93937770 on Build GRCh38
rs374433516 Chr.14: 93937737-93937737 on Build GRCh38
rs753414632 Chr.14: 93937807-93937807 on Build GRCh38
rs955849861 Chr.14: 93937834-93937834 on Build GRCh38
rs572067592 Chr.14: 93937839-93937839 on Build GRCh38
rs749921536 Chr.14: 93937754-93937754 on Build GRCh38
rs765782110 Chr.14: 93937811-93937811 on Build GRCh38
rs1031585530 Chr.14: 93937829-93937829 on Build GRCh38

View Full Product Details