Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49280452-49280677 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDADC1
Entrez Gene ID
81602
Gene Name
cytidine and dCMP deaminase domain containing 1
Gene Aliases
NYD-SP15, bA103J18.1
UniGene
Hs.388220
Gene Chromosome Location
Chr.13: 49247875-49293486 on Build GRCh38
Transcripts
NM_001193478.1 NM_030911.3 NR_036439.1 XM_006719871.3 XM_011535250.2 XM_011535251.1 XM_011535253.1 XM_017020778.1 XM_017020779.1 XM_017020780.1 XM_017020781.1 XM_017020782.1 XM_017020783.1 XM_017020784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6449445 c.1228G>C p.E410Q Chr.13: 49280516-49280516 on Build GRCh38
5950356 c.1333G>T p.D445Y Chr.13: 49280621-49280621 on Build GRCh38
256575 c.1323C>A p.I441I Chr.13: 49280611-49280611 on Build GRCh38
6227313 c.1255A>G p.I419V Chr.13: 49280543-49280543 on Build GRCh38
5030077 c.1331G>A p.G444E Chr.13: 49280619-49280619 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745544190 Chr.13: 49280565-49280565 on Build GRCh38
rs937367335 Chr.13: 49280608-49280608 on Build GRCh38
rs774328823 Chr.13: 49280624-49280624 on Build GRCh38
rs577964124 Chr.13: 49280499-49280499 on Build GRCh38
rs752110594 Chr.13: 49280532-49280532 on Build GRCh38
rs757756154 Chr.13: 49280539-49280539 on Build GRCh38
rs962503904 Chr.13: 49280554-49280554 on Build GRCh38
rs369535561 Chr.13: 49280488-49280488 on Build GRCh38
rs922276574 Chr.13: 49280580-49280580 on Build GRCh38
rs781455314 Chr.13: 49280533-49280535 on Build GRCh38
rs926531522 Chr.13: 49280605-49280605 on Build GRCh38
rs771861798 Chr.13: 49280630-49280630 on Build GRCh38
rs767283827 Chr.13: 49280505-49280505 on Build GRCh38
rs138682308 Chr.13: 49280509-49280509 on Build GRCh38
rs755673908 Chr.13: 49280594-49280594 on Build GRCh38
rs376230976 Chr.13: 49280596-49280596 on Build GRCh38
rs776855474 Chr.13: 49280506-49280506 on Build GRCh38
rs781634735 Chr.13: 49280540-49280540 on Build GRCh38
rs144640258 Chr.13: 49280507-49280507 on Build GRCh38
rs976356061 Chr.13: 49280555-49280555 on Build GRCh38
rs764549497 Chr.13: 49280514-49280514 on Build GRCh38
rs772955488 Chr.13: 49280643-49280643 on Build GRCh38
rs541137401 Chr.13: 49280496-49280497 on Build GRCh38
rs759344831 Chr.13: 49280487-49280491 on Build GRCh38
rs748786034 Chr.13: 49280599-49280599 on Build GRCh38
rs748299879 Chr.13: 49280560-49280560 on Build GRCh38
rs755049864 Chr.13: 49280529-49280532 on Build GRCh38
rs759829382 Chr.13: 49280651-49280651 on Build GRCh38
rs768203438 Chr.13: 49280615-49280615 on Build GRCh38
rs780128176 Chr.13: 49280497-49280497 on Build GRCh38
rs748017425 Chr.13: 49280627-49280627 on Build GRCh38

View Full Product Details