Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33085154-33085379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF3
Entrez Gene ID
1479
Gene Name
cleavage stimulation factor subunit 3
Gene Aliases
CSTF-77
UniGene
Hs.44402
Gene Chromosome Location
Chr.11: 33084584-33161491 on Build GRCh38
Transcripts
NM_001326.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5550318 c.1963G>A p.A655T Chr.11: 33085278-33085278 on Build GRCh38
6455540 c.1995G>A p.E665E Chr.11: 33085246-33085246 on Build GRCh38
926455 c.1952C>A p.T651N Chr.11: 33085289-33085289 on Build GRCh38
6571233 c.2083G>A p.E695K Chr.11: 33085158-33085158 on Build GRCh38
428924 c.2072C>T p.S691L Chr.11: 33085169-33085169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774690642 Chr.11: 33085260-33085260 on Build GRCh38
rs757337076 Chr.11: 33085221-33085221 on Build GRCh38
rs765652003 Chr.11: 33085295-33085299 on Build GRCh38
rs761919365 Chr.11: 33085339-33085339 on Build GRCh38
rs200568543 Chr.11: 33085228-33085228 on Build GRCh38
rs369236108 Chr.11: 33085326-33085326 on Build GRCh38
rs762715270 Chr.11: 33085297-33085297 on Build GRCh38
rs753943459 Chr.11: 33085206-33085206 on Build GRCh38
rs774293122 Chr.11: 33085312-33085312 on Build GRCh38
rs200163537 Chr.11: 33085313-33085313 on Build GRCh38
rs750644457 Chr.11: 33085223-33085223 on Build GRCh38
rs887562790 Chr.11: 33085302-33085302 on Build GRCh38
rs769147168 Chr.11: 33085259-33085259 on Build GRCh38
rs145202125 Chr.11: 33085255-33085255 on Build GRCh38
rs751491565 Chr.11: 33085191-33085191 on Build GRCh38
rs944760894 Chr.11: 33085346-33085346 on Build GRCh38
rs149939387 Chr.11: 33085192-33085192 on Build GRCh38
rs897324082 Chr.11: 33085320-33085320 on Build GRCh38
rs748152662 Chr.11: 33085342-33085342 on Build GRCh38
rs752622416 Chr.11: 33085338-33085338 on Build GRCh38
rs772878526 Chr.11: 33085293-33085293 on Build GRCh38
rs143330290 Chr.11: 33085321-33085322 on Build GRCh38
rs947718358 Chr.11: 33085194-33085194 on Build GRCh38
rs781609088 Chr.11: 33085262-33085262 on Build GRCh38
rs764009540 Chr.11: 33085201-33085201 on Build GRCh38
rs768753534 Chr.11: 33085240-33085240 on Build GRCh38
rs769229804 Chr.11: 33085273-33085273 on Build GRCh38
rs747673379 Chr.11: 33085271-33085271 on Build GRCh38
rs767472958 Chr.11: 33085327-33085327 on Build GRCh38
rs1005019430 Chr.11: 33085304-33085304 on Build GRCh38
rs35076726 Chr.11: 33085219-33085220 on Build GRCh38
rs770790627 Chr.11: 33085298-33085298 on Build GRCh38
rs779170010 Chr.11: 33085222-33085222 on Build GRCh38

View Full Product Details