Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 87407837-87408086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNH
Entrez Gene ID
902
Gene Name
cyclin H
Gene Aliases
CAK, CycH, p34, p37
UniGene
Hs.292524
Gene Chromosome Location
Chr.5: 87311480-87413033 on Build GRCh38
Transcripts
NM_001199189.1 NM_001239.3 XM_005248627.3 XM_005248629.4 XM_011543706.2 XM_017010014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618471 c.448C>T p.L150F Chr.5: 87408053-87408053 on Build GRCh38
1311372 c.491A>C p.Y164S Chr.5: 87408010-87408010 on Build GRCh38
221176 c.454C>A p.Q152K Chr.5: 87408047-87408047 on Build GRCh38
3140119 c.514A>G p.I172V Chr.5: 87407987-87407987 on Build GRCh38
1487022 c.421G>C p.E141Q Chr.5: 87408080-87408080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775837035 Chr.5: 87407988-87407988 on Build GRCh38
rs770037305 Chr.5: 87407984-87407984 on Build GRCh38
rs200767477 Chr.5: 87408011-87408011 on Build GRCh38
rs754752185 Chr.5: 87407931-87407931 on Build GRCh38
rs946031556 Chr.5: 87407876-87407876 on Build GRCh38
rs369841818 Chr.5: 87407997-87407997 on Build GRCh38
rs928352508 Chr.5: 87407857-87407857 on Build GRCh38
rs748053267 Chr.5: 87407945-87407945 on Build GRCh38
rs778602139 Chr.5: 87407938-87407938 on Build GRCh38
rs762293094 Chr.5: 87408000-87408000 on Build GRCh38
rs112808667 Chr.5: 87407883-87407883 on Build GRCh38
rs768003764 Chr.5: 87408003-87408003 on Build GRCh38
rs765894978 Chr.5: 87408020-87408020 on Build GRCh38
rs183348857 Chr.5: 87407906-87407906 on Build GRCh38
rs140164536 Chr.5: 87407924-87407924 on Build GRCh38
rs199815978 Chr.5: 87408037-87408037 on Build GRCh38
rs777915379 Chr.5: 87407950-87407950 on Build GRCh38
rs753489239 Chr.5: 87408021-87408021 on Build GRCh38
rs763108238 Chr.5: 87407994-87407994 on Build GRCh38
rs760155207 Chr.5: 87408014-87408014 on Build GRCh38
rs190762620 Chr.5: 87407877-87407877 on Build GRCh38
rs771919506 Chr.5: 87407946-87407946 on Build GRCh38
rs371442274 Chr.5: 87407881-87407881 on Build GRCh38
rs530898129 Chr.5: 87407860-87407860 on Build GRCh38
rs769027195 Chr.5: 87407996-87407996 on Build GRCh38
rs778714787 Chr.5: 87408049-87408049 on Build GRCh38
rs532625319 Chr.5: 87407885-87407885 on Build GRCh38
rs745938631 Chr.5: 87407952-87407952 on Build GRCh38

View Full Product Details