Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 147393938-147394211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL3
Entrez Gene ID
1809
Gene Name
dihydropyrimidinase like 3
Gene Aliases
CRMP-4, CRMP4, DRP-3, DRP3, LCRMP, ULIP, ULIP-1
UniGene
Hs.519659
Gene Chromosome Location
Chr.5: 147390808-147510056 on Build GRCh38
Transcripts
NM_001197294.1 NM_001387.2 XM_011537574.1 XM_011537577.2

Gene Symbol
STK32A
Entrez Gene ID
202374
Gene Name
serine/threonine kinase 32A
Gene Aliases
YANK1
UniGene
Hs.585069
Gene Chromosome Location
Chr.5: 147234963-147401996 on Build GRCh38
Transcripts
NM_001197294.1 NM_001387.2 XM_011537574.1 XM_011537577.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5809390 c.1625-2A>T p.? Chr.5: 147394125-147394125 on Build GRCh38
736492 c.1698C>G p.I566M Chr.5: 147394050-147394050 on Build GRCh38
1063906 c.1673C>T p.A558V Chr.5: 147394075-147394075 on Build GRCh38
3674350 c.1642G>T p.G548W Chr.5: 147394106-147394106 on Build GRCh38
5438998 c.1688G>T p.R563L Chr.5: 147394060-147394060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141439060 Chr.5: 147394170-147394171 on Build GRCh38
rs375392700 Chr.5: 147394064-147394064 on Build GRCh38
rs764061728 Chr.5: 147394146-147394146 on Build GRCh38
rs775308643 Chr.5: 147394019-147394020 on Build GRCh38
rs375957017 Chr.5: 147394171-147394171 on Build GRCh38
rs1025206541 Chr.5: 147394120-147394120 on Build GRCh38
rs17853868 Chr.5: 147394118-147394118 on Build GRCh38
rs998856637 Chr.5: 147394100-147394100 on Build GRCh38
rs778075066 Chr.5: 147394161-147394161 on Build GRCh38
rs751567310 Chr.5: 147394146-147394147 on Build GRCh38
rs1039611279 Chr.5: 147393968-147393968 on Build GRCh38
rs577467748 Chr.5: 147394059-147394059 on Build GRCh38
rs1033385957 Chr.5: 147394147-147394147 on Build GRCh38
rs200584034 Chr.5: 147394080-147394080 on Build GRCh38
rs35959663 Chr.5: 147394151-147394152 on Build GRCh38
rs909423927 Chr.5: 147393969-147393969 on Build GRCh38
rs773408854 Chr.5: 147394128-147394128 on Build GRCh38
rs759160130 Chr.5: 147393995-147393996 on Build GRCh38
rs774822283 Chr.5: 147394047-147394047 on Build GRCh38
rs760199898 Chr.5: 147394048-147394048 on Build GRCh38
rs763713401 Chr.5: 147394049-147394049 on Build GRCh38
rs756247241 Chr.5: 147394158-147394158 on Build GRCh38
rs542333524 Chr.5: 147394121-147394121 on Build GRCh38
rs3792837 Chr.5: 147394119-147394119 on Build GRCh38
rs756369537 Chr.5: 147394084-147394084 on Build GRCh38
rs745731115 Chr.5: 147393986-147393986 on Build GRCh38
rs759687251 Chr.5: 147394148-147394148 on Build GRCh38
rs182013586 Chr.5: 147394152-147394152 on Build GRCh38
rs763302616 Chr.5: 147394074-147394074 on Build GRCh38
rs771854528 Chr.5: 147394103-147394103 on Build GRCh38
rs752826138 Chr.5: 147394155-147394155 on Build GRCh38
rs933380784 Chr.5: 147394071-147394071 on Build GRCh38
rs921121964 Chr.5: 147393970-147393970 on Build GRCh38
rs940846847 Chr.5: 147394011-147394011 on Build GRCh38
rs769492854 Chr.5: 147394127-147394127 on Build GRCh38
rs780614068 Chr.5: 147394173-147394173 on Build GRCh38
rs1013612428 Chr.5: 147394124-147394124 on Build GRCh38
rs756308152 Chr.5: 147394161-147394162 on Build GRCh38
rs1051649933 Chr.5: 147394034-147394034 on Build GRCh38
rs987121988 Chr.5: 147394174-147394174 on Build GRCh38
rs762706222 Chr.5: 147394029-147394032 on Build GRCh38
rs562571897 Chr.5: 147394131-147394131 on Build GRCh38
rs531232275 Chr.5: 147394151-147394151 on Build GRCh38
rs778502005 Chr.5: 147394090-147394090 on Build GRCh38
rs564781290 Chr.5: 147394178-147394178 on Build GRCh38
rs780014551 Chr.5: 147394104-147394104 on Build GRCh38
rs3838662 Chr.5: 147394161-147394161 on Build GRCh38
rs764830950 Chr.5: 147394015-147394015 on Build GRCh38
rs758252014 Chr.5: 147394167-147394167 on Build GRCh38
rs936628864 Chr.5: 147394086-147394086 on Build GRCh38
rs376767463 Chr.5: 147394065-147394065 on Build GRCh38
rs766945584 Chr.5: 147394075-147394075 on Build GRCh38
rs750161505 Chr.5: 147394163-147394163 on Build GRCh38
rs752155595 Chr.5: 147394143-147394143 on Build GRCh38
rs386405242 Chr.5: 147394155-147394156 on Build GRCh38
rs755757436 Chr.5: 147394126-147394126 on Build GRCh38
rs779961117 Chr.5: 147394170-147394170 on Build GRCh38
rs773944258 Chr.5: 147394099-147394099 on Build GRCh38
rs768220433 Chr.5: 147394111-147394111 on Build GRCh38
rs371799829 Chr.5: 147394133-147394133 on Build GRCh38
rs374056960 Chr.5: 147394107-147394107 on Build GRCh38
rs772047096 Chr.5: 147394015-147394015 on Build GRCh38
rs778011131 Chr.5: 147394152-147394152 on Build GRCh38
rs1026447922 Chr.5: 147394154-147394154 on Build GRCh38
rs767602964 Chr.5: 147394156-147394156 on Build GRCh38
rs762162860 Chr.5: 147394060-147394060 on Build GRCh38
rs755161394 Chr.5: 147394079-147394079 on Build GRCh38
rs751645128 Chr.5: 147394077-147394077 on Build GRCh38

View Full Product Details