Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 67203437-67203710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAGAB
Entrez Gene ID
79719
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
KPPP1, PPKP1, PPKP1A, p34
UniGene
Hs.254642
Gene Chromosome Location
Chr.15: 67200675-67255198 on Build GRCh38
Transcripts
NM_001271885.1 NM_001271886.1 NM_024666.4 XM_011522020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589239 c.831G>A p.A277A Chr.15: 67203587-67203587 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1012043276 Chr.15: 67203540-67203540 on Build GRCh38
rs748533201 Chr.15: 67203576-67203576 on Build GRCh38
rs760255012 Chr.15: 67203535-67203535 on Build GRCh38
rs770736287 Chr.15: 67203513-67203513 on Build GRCh38
rs149079147 Chr.15: 67203475-67203475 on Build GRCh38
rs914620319 Chr.15: 67203604-67203604 on Build GRCh38
rs1025337499 Chr.15: 67203596-67203596 on Build GRCh38
rs780402119 Chr.15: 67203561-67203561 on Build GRCh38
rs202016609 Chr.15: 67203588-67203588 on Build GRCh38
rs756830257 Chr.15: 67203557-67203557 on Build GRCh38
rs763758300 Chr.15: 67203542-67203542 on Build GRCh38
rs767359660 Chr.15: 67203523-67203523 on Build GRCh38
rs746964492 Chr.15: 67203682-67203682 on Build GRCh38
rs759230701 Chr.15: 67203516-67203516 on Build GRCh38
rs868418659 Chr.15: 67203548-67203548 on Build GRCh38
rs898437068 Chr.15: 67203562-67203562 on Build GRCh38
rs114666833 Chr.15: 67203466-67203466 on Build GRCh38
rs1040305531 Chr.15: 67203681-67203681 on Build GRCh38
rs755388097 Chr.15: 67203571-67203571 on Build GRCh38
rs1003846421 Chr.15: 67203558-67203558 on Build GRCh38
rs944284632 Chr.15: 67203652-67203652 on Build GRCh38
rs536601770 Chr.15: 67203585-67203585 on Build GRCh38
rs760494051 Chr.15: 67203618-67203618 on Build GRCh38
rs368788098 Chr.15: 67203611-67203611 on Build GRCh38
rs749299363 Chr.15: 67203587-67203587 on Build GRCh38
rs533720832 Chr.15: 67203506-67203506 on Build GRCh38
rs759539510 Chr.15: 67203609-67203609 on Build GRCh38
rs754087987 Chr.15: 67203536-67203536 on Build GRCh38
rs774390658 Chr.15: 67203515-67203515 on Build GRCh38
rs568260692 Chr.15: 67203498-67203498 on Build GRCh38
rs975579895 Chr.15: 67203676-67203676 on Build GRCh38
rs554109653 Chr.15: 67203507-67203507 on Build GRCh38
rs752162972 Chr.15: 67203566-67203566 on Build GRCh38
rs912834824 Chr.15: 67203636-67203636 on Build GRCh38
rs770140551 Chr.15: 67203501-67203501 on Build GRCh38
rs753247583 Chr.15: 67203547-67203547 on Build GRCh38
rs779756552 Chr.15: 67203655-67203655 on Build GRCh38
rs138084549 Chr.15: 67203474-67203474 on Build GRCh38
rs772002801 Chr.15: 67203610-67203610 on Build GRCh38
rs776551074 Chr.15: 67203571-67203579 on Build GRCh38
rs763705280 Chr.15: 67203648-67203648 on Build GRCh38
rs964171892 Chr.15: 67203632-67203632 on Build GRCh38
rs781631892 Chr.15: 67203574-67203574 on Build GRCh38
rs752343070 Chr.15: 67203531-67203531 on Build GRCh38
rs774144509 Chr.15: 67203602-67203602 on Build GRCh38
rs770095615 Chr.15: 67203584-67203584 on Build GRCh38

View Full Product Details