Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114424341-114424576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WHRN
Entrez Gene ID
25861
Gene Name
whirlin
Gene Aliases
CIP98, DFNB31, PDZD7B, USH2D, WI
UniGene
Hs.93836
Gene Chromosome Location
Chr.9: 114402078-114505509 on Build GRCh38
Transcripts
NM_001083885.2 NM_001173425.1 NM_015404.3 XM_011518484.1 XM_011518485.1 XM_011518486.1 XM_011518487.1 XM_011518488.1 XM_011518489.2 XM_011518491.2 XM_011518492.1 XM_011518494.2 XM_011518495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4163264 c.1342G>A p.E448K Chr.9: 114424408-114424408 on Build GRCh38
6642832 c.246G>A p.L82L Chr.9: 114424451-114424451 on Build GRCh38
5767849 c.307G>A p.V103I Chr.9: 114424390-114424390 on Build GRCh38
4677135 c.255G>A p.E85E Chr.9: 114424442-114424442 on Build GRCh38
4677134 c.1308G>A p.E436E Chr.9: 114424442-114424442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111033459 Chr.9: 114424385-114424385 on Build GRCh38
rs749333910 Chr.9: 114424463-114424463 on Build GRCh38
rs200377723 Chr.9: 114424401-114424401 on Build GRCh38
rs770481636 Chr.9: 114424458-114424458 on Build GRCh38
rs397517256 Chr.9: 114424459-114424459 on Build GRCh38
rs759854771 Chr.9: 114424480-114424480 on Build GRCh38
rs201463786 Chr.9: 114424513-114424513 on Build GRCh38
rs778445696 Chr.9: 114424546-114424546 on Build GRCh38
rs745818473 Chr.9: 114424522-114424522 on Build GRCh38
rs748769354 Chr.9: 114424361-114424361 on Build GRCh38
rs757963300 Chr.9: 114424444-114424444 on Build GRCh38
rs370178817 Chr.9: 114424390-114424390 on Build GRCh38
rs201299182 Chr.9: 114424453-114424453 on Build GRCh38
rs748518724 Chr.9: 114424448-114424448 on Build GRCh38
rs756089219 Chr.9: 114424421-114424421 on Build GRCh38
rs778458791 Chr.9: 114424440-114424440 on Build GRCh38
rs774461811 Chr.9: 114424472-114424472 on Build GRCh38
rs753763480 Chr.9: 114424506-114424506 on Build GRCh38
rs117352600 Chr.9: 114424398-114424398 on Build GRCh38
rs756931477 Chr.9: 114424364-114424364 on Build GRCh38
rs35003670 Chr.9: 114424482-114424482 on Build GRCh38
rs889211270 Chr.9: 114424549-114424549 on Build GRCh38
rs145405120 Chr.9: 114424417-114424417 on Build GRCh38
rs4978584 Chr.9: 114424432-114424432 on Build GRCh38
rs149027634 Chr.9: 114424384-114424384 on Build GRCh38
rs745473400 Chr.9: 114424442-114424442 on Build GRCh38
rs373094583 Chr.9: 114424402-114424402 on Build GRCh38
rs148677152 Chr.9: 114424391-114424391 on Build GRCh38
rs760453597 Chr.9: 114424491-114424491 on Build GRCh38
rs769022650 Chr.9: 114424378-114424378 on Build GRCh38
rs1050377167 Chr.9: 114424533-114424533 on Build GRCh38
rs200905454 Chr.9: 114424516-114424516 on Build GRCh38
rs777297890 Chr.9: 114424379-114424379 on Build GRCh38
rs868739473 Chr.9: 114424462-114424462 on Build GRCh38
rs45530335 Chr.9: 114424419-114424419 on Build GRCh38
rs771004435 Chr.9: 114424464-114424464 on Build GRCh38
rs4979387 Chr.9: 114424397-114424397 on Build GRCh38
rs775334641 Chr.9: 114424490-114424490 on Build GRCh38
rs397517255 Chr.9: 114424483-114424483 on Build GRCh38
rs11539662 Chr.9: 114424423-114424423 on Build GRCh38
rs540699814 Chr.9: 114424360-114424360 on Build GRCh38
rs761263741 Chr.9: 114424508-114424508 on Build GRCh38
rs758129253 Chr.9: 114424523-114424523 on Build GRCh38
rs777908643 Chr.9: 114424416-114424416 on Build GRCh38
rs376346064 Chr.9: 114424426-114424426 on Build GRCh38
rs150407952 Chr.9: 114424445-114424445 on Build GRCh38
rs753116383 Chr.9: 114424536-114424536 on Build GRCh38
rs373389359 Chr.9: 114424372-114424372 on Build GRCh38
rs527784694 Chr.9: 114424493-114424493 on Build GRCh38
rs764019960 Chr.9: 114424494-114424494 on Build GRCh38
rs144878400 Chr.9: 114424369-114424369 on Build GRCh38
rs886063372 Chr.9: 114424399-114424399 on Build GRCh38
rs778565930 Chr.9: 114424365-114424365 on Build GRCh38
rs368241259 Chr.9: 114424528-114424528 on Build GRCh38
rs781184468 Chr.9: 114424434-114424434 on Build GRCh38
rs756715564 Chr.9: 114424433-114424433 on Build GRCh38

View Full Product Details