Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75651448-75651721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C14orf1
Entrez Gene ID
11161
Gene Name
chromosome 14 open reading frame 1
Gene Aliases
ERG28, NET51
UniGene
Hs.15106
Gene Chromosome Location
Chr.14: 75650890-75661195 on Build GRCh38
Transcripts
NM_007176.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4938894 c.363G>A p.M121I Chr.14: 75651615-75651615 on Build GRCh38
1735383 c.361A>T p.M121L Chr.14: 75651617-75651617 on Build GRCh38
1371179 c.411G>T p.K137N Chr.14: 75651567-75651567 on Build GRCh38
4491171 c.375C>T p.L125L Chr.14: 75651603-75651603 on Build GRCh38
1198043 c.376C>T p.R126W Chr.14: 75651602-75651602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567266543 Chr.14: 75651587-75651587 on Build GRCh38
rs369318513 Chr.14: 75651579-75651579 on Build GRCh38
rs114809316 Chr.14: 75651593-75651593 on Build GRCh38
rs1057065264 Chr.14: 75651484-75651484 on Build GRCh38
rs745626669 Chr.14: 75651614-75651614 on Build GRCh38
rs770906607 Chr.14: 75651508-75651508 on Build GRCh38
rs919416185 Chr.14: 75651627-75651627 on Build GRCh38
rs764778066 Chr.14: 75651584-75651584 on Build GRCh38
rs376262367 Chr.14: 75651525-75651525 on Build GRCh38
rs997217926 Chr.14: 75651495-75651495 on Build GRCh38
rs780418393 Chr.14: 75651645-75651645 on Build GRCh38
rs183825619 Chr.14: 75651581-75651581 on Build GRCh38
rs985824314 Chr.14: 75651539-75651539 on Build GRCh38
rs902160436 Chr.14: 75651573-75651573 on Build GRCh38
rs754068250 Chr.14: 75651518-75651518 on Build GRCh38
rs757095170 Chr.14: 75651608-75651608 on Build GRCh38
rs775703018 Chr.14: 75651676-75651677 on Build GRCh38
rs903060214 Chr.14: 75651492-75651492 on Build GRCh38
rs530426881 Chr.14: 75651547-75651547 on Build GRCh38
rs372857013 Chr.14: 75651515-75651515 on Build GRCh38
rs922850717 Chr.14: 75651493-75651493 on Build GRCh38
rs946440030 Chr.14: 75651549-75651549 on Build GRCh38
rs776233515 Chr.14: 75651512-75651512 on Build GRCh38
rs373496095 Chr.14: 75651649-75651649 on Build GRCh38
rs781227300 Chr.14: 75651609-75651609 on Build GRCh38
rs953146449 Chr.14: 75651530-75651530 on Build GRCh38
rs188419927 Chr.14: 75651680-75651680 on Build GRCh38
rs761639946 Chr.14: 75651684-75651685 on Build GRCh38
rs751084984 Chr.14: 75651602-75651602 on Build GRCh38
rs764320361 Chr.14: 75651496-75651496 on Build GRCh38
rs115877512 Chr.14: 75651572-75651572 on Build GRCh38
rs777198821 Chr.14: 75651601-75651601 on Build GRCh38
rs1012581212 Chr.14: 75651674-75651674 on Build GRCh38
rs763111662 Chr.14: 75651528-75651528 on Build GRCh38
rs774573485 Chr.14: 75651560-75651560 on Build GRCh38
rs982179790 Chr.14: 75651681-75651681 on Build GRCh38
rs769874675 Chr.14: 75651524-75651524 on Build GRCh38
rs762154723 Chr.14: 75651617-75651617 on Build GRCh38
rs376784047 Chr.14: 75651673-75651673 on Build GRCh38

View Full Product Details