Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 105524306-105524565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALCAM
Entrez Gene ID
214
Gene Name
activated leukocyte cell adhesion molecule
Gene Aliases
CD166, MEMD
UniGene
Hs.591293
Gene Chromosome Location
Chr.3: 105366713-105576913 on Build GRCh38
Transcripts
NM_001243280.1 NM_001243281.1 NM_001243283.1 NM_001627.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035935 c.202A>G p.I68V Chr.3: 105524316-105524316 on Build GRCh38
3780616 c.212G>C p.R71T Chr.3: 105524326-105524326 on Build GRCh38
342449 c.329G>A p.R110K Chr.3: 105524443-105524443 on Build GRCh38
4660980 c.388G>T p.V130L Chr.3: 105524502-105524502 on Build GRCh38
3845721 c.322G>T p.E108* Chr.3: 105524436-105524436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753780904 Chr.3: 105524474-105524474 on Build GRCh38
rs747372908 Chr.3: 105524333-105524333 on Build GRCh38
rs746416911 Chr.3: 105524344-105524344 on Build GRCh38
rs751744021 Chr.3: 105524487-105524487 on Build GRCh38
rs976285561 Chr.3: 105524363-105524363 on Build GRCh38
rs762442983 Chr.3: 105524355-105524355 on Build GRCh38
rs368056953 Chr.3: 105524466-105524466 on Build GRCh38
rs147777853 Chr.3: 105524357-105524357 on Build GRCh38
rs769133849 Chr.3: 105524347-105524347 on Build GRCh38
rs775077113 Chr.3: 105524354-105524354 on Build GRCh38
rs368193586 Chr.3: 105524358-105524358 on Build GRCh38
rs761587401 Chr.3: 105524425-105524425 on Build GRCh38
rs953822868 Chr.3: 105524431-105524431 on Build GRCh38
rs776933959 Chr.3: 105524456-105524467 on Build GRCh38
rs768329672 Chr.3: 105524408-105524408 on Build GRCh38
rs748817733 Chr.3: 105524404-105524404 on Build GRCh38
rs760463121 Chr.3: 105524462-105524462 on Build GRCh38
rs752827223 Chr.3: 105524369-105524369 on Build GRCh38
rs771346909 Chr.3: 105524341-105524341 on Build GRCh38
rs755883922 Chr.3: 105524439-105524439 on Build GRCh38
rs377050023 Chr.3: 105524475-105524475 on Build GRCh38
rs929797653 Chr.3: 105524521-105524521 on Build GRCh38
rs577291146 Chr.3: 105524534-105524534 on Build GRCh38
rs748382272 Chr.3: 105524481-105524481 on Build GRCh38
rs780488374 Chr.3: 105524400-105524400 on Build GRCh38
rs750792996 Chr.3: 105524504-105524504 on Build GRCh38
rs768241426 Chr.3: 105524540-105524540 on Build GRCh38
rs756490581 Chr.3: 105524520-105524520 on Build GRCh38
rs537535274 Chr.3: 105524412-105524412 on Build GRCh38
rs371134471 Chr.3: 105524542-105524542 on Build GRCh38
rs149366678 Chr.3: 105524386-105524386 on Build GRCh38
rs781465503 Chr.3: 105524502-105524502 on Build GRCh38
rs752485754 Chr.3: 105524388-105524388 on Build GRCh38
rs150607806 Chr.3: 105524360-105524360 on Build GRCh38
rs1045575560 Chr.3: 105524529-105524529 on Build GRCh38
rs749797146 Chr.3: 105524537-105524537 on Build GRCh38
rs558809274 Chr.3: 105524478-105524478 on Build GRCh38
rs777963062 Chr.3: 105524382-105524382 on Build GRCh38
rs191774794 Chr.3: 105524383-105524383 on Build GRCh38
rs918535348 Chr.3: 105524491-105524491 on Build GRCh38
rs770334153 Chr.3: 105524396-105524396 on Build GRCh38
rs758474954 Chr.3: 105524373-105524373 on Build GRCh38
rs757481160 Chr.3: 105524497-105524497 on Build GRCh38
rs144550658 Chr.3: 105524394-105524394 on Build GRCh38
rs9875739 Chr.3: 105524545-105524545 on Build GRCh38
rs141090189 Chr.3: 105524365-105524365 on Build GRCh38
rs777102845 Chr.3: 105524343-105524343 on Build GRCh38
rs759732563 Chr.3: 105524359-105524359 on Build GRCh38
rs765175016 Chr.3: 105524367-105524367 on Build GRCh38
rs771945333 Chr.3: 105524432-105524432 on Build GRCh38

View Full Product Details