Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45034266-45034505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAKD
Entrez Gene ID
79877
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
UniGene
Hs.463148
Gene Chromosome Location
Chr.17: 45023338-45061129 on Build GRCh38
Transcripts
NM_001128631.2 NM_001288654.2 NM_001288655.1 NM_001321326.1 NM_024819.6 XM_005257688.2 XM_011525262.2 XM_017025102.1 XM_017025103.1 XM_017025104.1 XM_017025105.1 XM_017025106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972227 c.159C>T p.F53F Chr.17: 45034344-45034344 on Build GRCh38
5972225 c.213C>T p.D71D Chr.17: 45034290-45034290 on Build GRCh38
1383761 c.196C>T p.R66C Chr.17: 45034307-45034307 on Build GRCh38
6146929 c.154G>T p.V52F Chr.17: 45034349-45034349 on Build GRCh38
1383760 c.230C>A p.P77H Chr.17: 45034273-45034273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777282741 Chr.17: 45034319-45034320 on Build GRCh38
rs750401195 Chr.17: 45034338-45034338 on Build GRCh38
rs751840593 Chr.17: 45034381-45034381 on Build GRCh38
rs374062073 Chr.17: 45034435-45034435 on Build GRCh38
rs181186960 Chr.17: 45034296-45034296 on Build GRCh38
rs780810927 Chr.17: 45034335-45034335 on Build GRCh38
rs764385345 Chr.17: 45034319-45034319 on Build GRCh38
rs779979696 Chr.17: 45034396-45034396 on Build GRCh38
rs770257411 Chr.17: 45034428-45034428 on Build GRCh38
rs974172129 Chr.17: 45034344-45034344 on Build GRCh38
rs141495277 Chr.17: 45034363-45034363 on Build GRCh38
rs746243972 Chr.17: 45034431-45034431 on Build GRCh38
rs774958081 Chr.17: 45034290-45034290 on Build GRCh38
rs754543676 Chr.17: 45034400-45034400 on Build GRCh38
rs766601603 Chr.17: 45034306-45034306 on Build GRCh38
rs747881153 Chr.17: 45034418-45034418 on Build GRCh38
rs199996338 Chr.17: 45034361-45034361 on Build GRCh38
rs764031953 Chr.17: 45034379-45034379 on Build GRCh38
rs371564171 Chr.17: 45034411-45034411 on Build GRCh38
rs199654954 Chr.17: 45034317-45034317 on Build GRCh38
rs776126345 Chr.17: 45034403-45034405 on Build GRCh38
rs769325051 Chr.17: 45034286-45034286 on Build GRCh38
rs761111925 Chr.17: 45034303-45034303 on Build GRCh38
rs150715564 Chr.17: 45034389-45034389 on Build GRCh38
rs763140090 Chr.17: 45034364-45034364 on Build GRCh38
rs779603355 Chr.17: 45034341-45034341 on Build GRCh38
rs763336081 Chr.17: 45034318-45034318 on Build GRCh38
rs143735388 Chr.17: 45034316-45034316 on Build GRCh38
rs772566334 Chr.17: 45034372-45034372 on Build GRCh38
rs771364324 Chr.17: 45034358-45034358 on Build GRCh38
rs750212915 Chr.17: 45034394-45034394 on Build GRCh38
rs1032154831 Chr.17: 45034479-45034479 on Build GRCh38
rs142968159 Chr.17: 45034325-45034325 on Build GRCh38
rs1051466382 Chr.17: 45034482-45034482 on Build GRCh38
rs374249436 Chr.17: 45034355-45034355 on Build GRCh38
rs767992972 Chr.17: 45034351-45034351 on Build GRCh38
rs954861234 Chr.17: 45034308-45034308 on Build GRCh38
rs368505042 Chr.17: 45034397-45034397 on Build GRCh38
rs547534311 Chr.17: 45034483-45034483 on Build GRCh38
rs748998128 Chr.17: 45034292-45034292 on Build GRCh38
rs900599946 Chr.17: 45034484-45034484 on Build GRCh38
rs377710350 Chr.17: 45034313-45034313 on Build GRCh38
rs9900173 Chr.17: 45034320-45034320 on Build GRCh38
rs1052236595 Chr.17: 45034424-45034424 on Build GRCh38
rs139028901 Chr.17: 45034360-45034360 on Build GRCh38
rs371641816 Chr.17: 45034334-45034334 on Build GRCh38
rs777222686 Chr.17: 45034307-45034307 on Build GRCh38
rs565333243 Chr.17: 45034434-45034434 on Build GRCh38
rs760084683 Chr.17: 45034309-45034309 on Build GRCh38
rs913668981 Chr.17: 45034449-45034449 on Build GRCh38
rs747710663 Chr.17: 45034356-45034356 on Build GRCh38
rs748841150 Chr.17: 45034320-45034321 on Build GRCh38
rs201742887 Chr.17: 45034388-45034388 on Build GRCh38
rs145138207 Chr.17: 45034343-45034343 on Build GRCh38
rs184085169 Chr.17: 45034473-45034473 on Build GRCh38
rs756031085 Chr.17: 45034340-45034340 on Build GRCh38

View Full Product Details