Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34456598-34456869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAT
Entrez Gene ID
847
Gene Name
catalase
Gene Aliases
-
UniGene
Hs.502302
Gene Chromosome Location
Chr.11: 34438925-34472060 on Build GRCh38
Transcripts
NM_001752.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700061 c.983A>G p.E328G Chr.11: 34456744-34456744 on Build GRCh38
926631 c.1003G>A p.D335N Chr.11: 34456764-34456764 on Build GRCh38
5838470 c.959G>A p.R320Q Chr.11: 34456720-34456720 on Build GRCh38
4032531 c.1000T>G p.F334V Chr.11: 34456761-34456761 on Build GRCh38
289431 c.1047A>T p.K349N Chr.11: 34456808-34456808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760627725 Chr.11: 34456695-34456706 on Build GRCh38
rs141526214 Chr.11: 34456671-34456671 on Build GRCh38
rs770822179 Chr.11: 34456733-34456737 on Build GRCh38
rs751811203 Chr.11: 34456719-34456719 on Build GRCh38
rs759821907 Chr.11: 34456837-34456837 on Build GRCh38
rs139421991 Chr.11: 34456720-34456720 on Build GRCh38
rs370314175 Chr.11: 34456703-34456703 on Build GRCh38
rs551303235 Chr.11: 34456800-34456800 on Build GRCh38
rs773794289 Chr.11: 34456647-34456648 on Build GRCh38
rs777127586 Chr.11: 34456763-34456763 on Build GRCh38
rs748703348 Chr.11: 34456674-34456674 on Build GRCh38
rs745533600 Chr.11: 34456783-34456783 on Build GRCh38
rs751715058 Chr.11: 34456832-34456832 on Build GRCh38
rs772847602 Chr.11: 34456676-34456676 on Build GRCh38
rs766469822 Chr.11: 34456830-34456830 on Build GRCh38
rs758595438 Chr.11: 34456709-34456709 on Build GRCh38
rs571047357 Chr.11: 34456818-34456818 on Build GRCh38
rs774258742 Chr.11: 34456686-34456686 on Build GRCh38
rs781624442 Chr.11: 34456732-34456732 on Build GRCh38
rs371223871 Chr.11: 34456647-34456647 on Build GRCh38
rs748936117 Chr.11: 34456682-34456682 on Build GRCh38
rs921909672 Chr.11: 34456643-34456643 on Build GRCh38
rs768685531 Chr.11: 34456801-34456801 on Build GRCh38
rs770598518 Chr.11: 34456774-34456774 on Build GRCh38
rs770759609 Chr.11: 34456684-34456684 on Build GRCh38
rs367899200 Chr.11: 34456764-34456764 on Build GRCh38
rs756704996 Chr.11: 34456760-34456760 on Build GRCh38
rs150895196 Chr.11: 34456691-34456691 on Build GRCh38
rs771692938 Chr.11: 34456798-34456798 on Build GRCh38
rs753141249 Chr.11: 34456753-34456753 on Build GRCh38
rs781511055 Chr.11: 34456672-34456672 on Build GRCh38
rs956323304 Chr.11: 34456688-34456688 on Build GRCh38
rs760662268 Chr.11: 34456699-34456699 on Build GRCh38
rs200381065 Chr.11: 34456675-34456675 on Build GRCh38
rs199789645 Chr.11: 34456841-34456841 on Build GRCh38
rs767761717 Chr.11: 34456843-34456843 on Build GRCh38
rs199818445 Chr.11: 34456708-34456708 on Build GRCh38
rs767446024 Chr.11: 34456687-34456687 on Build GRCh38
rs762979529 Chr.11: 34456820-34456820 on Build GRCh38
rs778664352 Chr.11: 34456782-34456782 on Build GRCh38
rs766625891 Chr.11: 34456710-34456710 on Build GRCh38
rs375479206 Chr.11: 34456650-34456650 on Build GRCh38

View Full Product Details