Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 214780428-214780701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BARD1
Entrez Gene ID
580
Gene Name
BRCA1 associated RING domain 1
Gene Aliases
-
UniGene
Hs.591642
Gene Chromosome Location
Chr.2: 214725645-214809711 on Build GRCh38
Transcripts
NM_000465.3 NM_001282543.1 NM_001282545.1 NM_001282548.1 NM_001282549.1 NR_104212.1 NR_104215.1 NR_104216.1 XM_017004613.1 XM_017004614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3577418 c.1304C>T p.A435V Chr.2: 214780570-214780570 on Build GRCh38
5497692 c.1219G>A p.V407M Chr.2: 214780655-214780655 on Build GRCh38
5892696 c.1265T>G p.V422G Chr.2: 214780609-214780609 on Build GRCh38
720080 c.1232C>A p.P411H Chr.2: 214780642-214780642 on Build GRCh38
720081 c.1237G>A p.A413T Chr.2: 214780637-214780637 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115533621 Chr.2: 214780523-214780523 on Build GRCh38
rs371409193 Chr.2: 214780609-214780610 on Build GRCh38
rs786202226 Chr.2: 214780648-214780648 on Build GRCh38
rs532375233 Chr.2: 214780494-214780494 on Build GRCh38
rs587780015 Chr.2: 214780585-214780585 on Build GRCh38
rs876659789 Chr.2: 214780563-214780563 on Build GRCh38
rs372679962 Chr.2: 214780569-214780569 on Build GRCh38
rs796666047 Chr.2: 214780669-214780669 on Build GRCh38
rs764364930 Chr.2: 214780535-214780535 on Build GRCh38
rs185164724 Chr.2: 214780580-214780580 on Build GRCh38
rs1049644277 Chr.2: 214780466-214780466 on Build GRCh38
rs759637650 Chr.2: 214780515-214780515 on Build GRCh38
rs371830908 Chr.2: 214780547-214780547 on Build GRCh38
rs587780014 Chr.2: 214780657-214780657 on Build GRCh38
rs192972931 Chr.2: 214780549-214780549 on Build GRCh38
rs76824305 Chr.2: 214780609-214780609 on Build GRCh38
rs786203605 Chr.2: 214780566-214780566 on Build GRCh38
rs757900534 Chr.2: 214780656-214780656 on Build GRCh38
rs188181984 Chr.2: 214780481-214780481 on Build GRCh38
rs902123015 Chr.2: 214780501-214780501 on Build GRCh38
rs765212498 Chr.2: 214780635-214780635 on Build GRCh38
rs370553043 Chr.2: 214780671-214780671 on Build GRCh38
rs889203476 Chr.2: 214780472-214780472 on Build GRCh38
rs876659775 Chr.2: 214780600-214780600 on Build GRCh38
rs786203499 Chr.2: 214780598-214780598 on Build GRCh38
rs876660910 Chr.2: 214780610-214780610 on Build GRCh38
rs943563066 Chr.2: 214780482-214780482 on Build GRCh38
rs876660911 Chr.2: 214780604-214780604 on Build GRCh38
rs876659685 Chr.2: 214780674-214780674 on Build GRCh38
rs786203533 Chr.2: 214780607-214780610 on Build GRCh38
rs537611647 Chr.2: 214780587-214780587 on Build GRCh38
rs754539312 Chr.2: 214780562-214780562 on Build GRCh38
rs786203825 Chr.2: 214780643-214780643 on Build GRCh38
rs781482219 Chr.2: 214780680-214780680 on Build GRCh38
rs587782681 Chr.2: 214780662-214780662 on Build GRCh38
rs786203162 Chr.2: 214780668-214780668 on Build GRCh38
rs753785671 Chr.2: 214780559-214780559 on Build GRCh38
rs199834790 Chr.2: 214780619-214780619 on Build GRCh38
rs766209380 Chr.2: 214780612-214780612 on Build GRCh38
rs749383704 Chr.2: 214780606-214780606 on Build GRCh38
rs876660778 Chr.2: 214780639-214780639 on Build GRCh38
rs876660639 Chr.2: 214780594-214780594 on Build GRCh38
rs759601398 Chr.2: 214780634-214780634 on Build GRCh38
rs786203217 Chr.2: 214780615-214780615 on Build GRCh38
rs868089048 Chr.2: 214780555-214780555 on Build GRCh38
rs878853996 Chr.2: 214780627-214780627 on Build GRCh38
rs1042167863 Chr.2: 214780457-214780457 on Build GRCh38
rs764592698 Chr.2: 214780565-214780565 on Build GRCh38
rs377153250 Chr.2: 214780658-214780658 on Build GRCh38
rs878853997 Chr.2: 214780589-214780591 on Build GRCh38
rs201729742 Chr.2: 214780679-214780679 on Build GRCh38
rs752234478 Chr.2: 214780567-214780567 on Build GRCh38
rs867396444 Chr.2: 214780676-214780676 on Build GRCh38
rs879254117 Chr.2: 214780546-214780546 on Build GRCh38
rs878853998 Chr.2: 214780589-214780589 on Build GRCh38
rs547346616 Chr.2: 214780517-214780517 on Build GRCh38
rs201146688 Chr.2: 214780607-214780607 on Build GRCh38
rs772950179 Chr.2: 214780545-214780545 on Build GRCh38
rs758049210 Chr.2: 214780574-214780574 on Build GRCh38
rs879254118 Chr.2: 214780630-214780630 on Build GRCh38
rs752186689 Chr.2: 214780641-214780641 on Build GRCh38
rs999023601 Chr.2: 214780522-214780522 on Build GRCh38
rs141351703 Chr.2: 214780454-214780461 on Build GRCh38
rs775490960 Chr.2: 214780605-214780605 on Build GRCh38
rs71579845 Chr.2: 214780531-214780531 on Build GRCh38
rs786203891 Chr.2: 214780676-214780676 on Build GRCh38
rs587781970 Chr.2: 214780576-214780576 on Build GRCh38
rs879253879 Chr.2: 214780590-214780590 on Build GRCh38
rs876659886 Chr.2: 214780661-214780661 on Build GRCh38
rs776723079 Chr.2: 214780618-214780618 on Build GRCh38
rs936401274 Chr.2: 214780476-214780476 on Build GRCh38

View Full Product Details