Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67457524-67457751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP4
Entrez Gene ID
57010
Gene Name
calcium binding protein 4
Gene Aliases
CRSD, CSNB2B
UniGene
Hs.143036
Gene Chromosome Location
Chr.11: 67452406-67461774 on Build GRCh38
Transcripts
NM_001300895.1 NM_001300896.1 NM_145200.3 XM_005274114.3 XM_011545181.2 XM_011545182.2 XM_011545183.2 XM_017018025.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3383770 c.612C>A p.H204Q Chr.11: 67457643-67457643 on Build GRCh38
4668283 c.651G>A p.E217E Chr.11: 67457682-67457682 on Build GRCh38
931096 c.635G>A p.R212H Chr.11: 67457666-67457666 on Build GRCh38
4668284 c.336G>A p.E112E Chr.11: 67457682-67457682 on Build GRCh38
3398077 c.310C>T p.R104* Chr.11: 67457656-67457656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752961611 Chr.11: 67457628-67457628 on Build GRCh38
rs150929930 Chr.11: 67457636-67457636 on Build GRCh38
rs777888510 Chr.11: 67457577-67457577 on Build GRCh38
rs142530984 Chr.11: 67457629-67457629 on Build GRCh38
rs755673472 Chr.11: 67457728-67457728 on Build GRCh38
rs779788706 Chr.11: 67457656-67457656 on Build GRCh38
rs766416524 Chr.11: 67457603-67457603 on Build GRCh38
rs774175876 Chr.11: 67457665-67457665 on Build GRCh38
rs775087763 Chr.11: 67457582-67457582 on Build GRCh38
rs1025537529 Chr.11: 67457706-67457706 on Build GRCh38
rs764548658 Chr.11: 67457696-67457696 on Build GRCh38
rs765512601 Chr.11: 67457618-67457618 on Build GRCh38
rs768544037 Chr.11: 67457589-67457589 on Build GRCh38
rs146764702 Chr.11: 67457578-67457578 on Build GRCh38
rs749115440 Chr.11: 67457585-67457585 on Build GRCh38
rs765324439 Chr.11: 67457678-67457678 on Build GRCh38
rs373263875 Chr.11: 67457666-67457666 on Build GRCh38
rs377174577 Chr.11: 67457687-67457687 on Build GRCh38
rs778453952 Chr.11: 67457566-67457566 on Build GRCh38
rs139392451 Chr.11: 67457610-67457610 on Build GRCh38
rs199749736 Chr.11: 67457697-67457697 on Build GRCh38
rs550529214 Chr.11: 67457730-67457730 on Build GRCh38
rs989681114 Chr.11: 67457557-67457557 on Build GRCh38
rs1016513036 Chr.11: 67457715-67457715 on Build GRCh38
rs772058303 Chr.11: 67457670-67457670 on Build GRCh38
rs200515541 Chr.11: 67457650-67457650 on Build GRCh38
rs749861593 Chr.11: 67457720-67457720 on Build GRCh38
rs774298352 Chr.11: 67457597-67457597 on Build GRCh38
rs892508675 Chr.11: 67457692-67457692 on Build GRCh38
rs369700438 Chr.11: 67457581-67457581 on Build GRCh38
rs552701854 Chr.11: 67457657-67457657 on Build GRCh38
rs150115958 Chr.11: 67457677-67457677 on Build GRCh38
rs745394874 Chr.11: 67457640-67457640 on Build GRCh38
rs745499827 Chr.11: 67457580-67457580 on Build GRCh38
rs781141438 Chr.11: 67457702-67457711 on Build GRCh38
rs1051333994 Chr.11: 67457644-67457644 on Build GRCh38
rs757905556 Chr.11: 67457637-67457637 on Build GRCh38
rs140655456 Chr.11: 67457639-67457639 on Build GRCh38
rs776838525 Chr.11: 67457609-67457609 on Build GRCh38
rs761792228 Chr.11: 67457602-67457602 on Build GRCh38
rs1055218639 Chr.11: 67457613-67457613 on Build GRCh38
rs890965729 Chr.11: 67457653-67457653 on Build GRCh38
rs757779036 Chr.11: 67457707-67457707 on Build GRCh38
rs758906973 Chr.11: 67457564-67457564 on Build GRCh38
rs897094997 Chr.11: 67457648-67457648 on Build GRCh38
rs969564762 Chr.11: 67457708-67457709 on Build GRCh38
rs764599417 Chr.11: 67457635-67457635 on Build GRCh38
rs1038691478 Chr.11: 67457645-67457645 on Build GRCh38
rs763303147 Chr.11: 67457695-67457695 on Build GRCh38
rs201059902 Chr.11: 67457710-67457710 on Build GRCh38

View Full Product Details