Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7562587-7562856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSP
Entrez Gene ID
1832
Gene Name
desmoplakin
Gene Aliases
DCWHKTA, DP
UniGene
Hs.519873
Gene Chromosome Location
Chr.6: 7541575-7586717 on Build GRCh38
Transcripts
NM_001008844.2 NM_001319034.1 NM_004415.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5632777 c.692A>G p.Y231C Chr.6: 7562746-7562746 on Build GRCh38
6649651 c.599C>T p.A200V Chr.6: 7562653-7562653 on Build GRCh38
7276468 c.603G>A p.E201E Chr.6: 7562657-7562657 on Build GRCh38
4625844 c.648G>A p.Q216Q Chr.6: 7562702-7562702 on Build GRCh38
274888 c.629A>G p.D210G Chr.6: 7562683-7562683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777091556 Chr.6: 7562651-7562651 on Build GRCh38
rs755724098 Chr.6: 7562616-7562616 on Build GRCh38
rs748400605 Chr.6: 7562654-7562654 on Build GRCh38
rs376185352 Chr.6: 7562739-7562739 on Build GRCh38
rs367986893 Chr.6: 7562831-7562831 on Build GRCh38
rs774450044 Chr.6: 7562784-7562784 on Build GRCh38
rs940238260 Chr.6: 7562770-7562770 on Build GRCh38
rs397516956 Chr.6: 7562766-7562767 on Build GRCh38
rs755544573 Chr.6: 7562827-7562827 on Build GRCh38
rs780207295 Chr.6: 7562679-7562679 on Build GRCh38
rs1044634464 Chr.6: 7562760-7562760 on Build GRCh38
rs770210088 Chr.6: 7562746-7562746 on Build GRCh38
rs763647233 Chr.6: 7562724-7562724 on Build GRCh38
rs749438581 Chr.6: 7562749-7562749 on Build GRCh38
rs375069870 Chr.6: 7562631-7562631 on Build GRCh38
rs753444512 Chr.6: 7562738-7562738 on Build GRCh38
rs760447383 Chr.6: 7562813-7562813 on Build GRCh38
rs794728109 Chr.6: 7562670-7562670 on Build GRCh38
rs1002592589 Chr.6: 7562610-7562610 on Build GRCh38
rs397516955 Chr.6: 7562753-7562753 on Build GRCh38
rs934038992 Chr.6: 7562736-7562736 on Build GRCh38
rs868293429 Chr.6: 7562775-7562775 on Build GRCh38
rs373862318 Chr.6: 7562626-7562626 on Build GRCh38
rs927601472 Chr.6: 7562790-7562790 on Build GRCh38
rs112877191 Chr.6: 7562741-7562741 on Build GRCh38
rs770080985 Chr.6: 7562657-7562657 on Build GRCh38
rs758845916 Chr.6: 7562634-7562635 on Build GRCh38
rs777967958 Chr.6: 7562831-7562840 on Build GRCh38
rs550684292 Chr.6: 7562832-7562832 on Build GRCh38
rs370309952 Chr.6: 7562787-7562787 on Build GRCh38
rs763855095 Chr.6: 7562822-7562822 on Build GRCh38
rs774136644 Chr.6: 7562699-7562699 on Build GRCh38
rs1028793473 Chr.6: 7562688-7562688 on Build GRCh38
rs765653540 Chr.6: 7562647-7562647 on Build GRCh38
rs147315869 Chr.6: 7562742-7562742 on Build GRCh38
rs761482224 Chr.6: 7562824-7562824 on Build GRCh38
rs762939968 Chr.6: 7562694-7562694 on Build GRCh38
rs759008993 Chr.6: 7562633-7562633 on Build GRCh38
rs987363401 Chr.6: 7562706-7562706 on Build GRCh38
rs374100807 Chr.6: 7562661-7562661 on Build GRCh38
rs148095061 Chr.6: 7562697-7562697 on Build GRCh38
rs199509943 Chr.6: 7562722-7562722 on Build GRCh38
rs546805348 Chr.6: 7562740-7562740 on Build GRCh38
rs752845209 Chr.6: 7562617-7562617 on Build GRCh38
rs111295218 Chr.6: 7562781-7562781 on Build GRCh38
rs375407471 Chr.6: 7562730-7562730 on Build GRCh38
rs767321201 Chr.6: 7562659-7562659 on Build GRCh38
rs777847026 Chr.6: 7562748-7562748 on Build GRCh38
rs775269539 Chr.6: 7562800-7562800 on Build GRCh38
rs996098764 Chr.6: 7562681-7562681 on Build GRCh38
rs113478603 Chr.6: 7562782-7562782 on Build GRCh38
rs1021314728 Chr.6: 7562666-7562666 on Build GRCh38
rs200806163 Chr.6: 7562721-7562721 on Build GRCh38
rs182623208 Chr.6: 7562823-7562823 on Build GRCh38
rs747448946 Chr.6: 7562648-7562648 on Build GRCh38
rs769107659 Chr.6: 7562650-7562650 on Build GRCh38
rs921332894 Chr.6: 7562731-7562731 on Build GRCh38
rs377749481 Chr.6: 7562719-7562719 on Build GRCh38

View Full Product Details