Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15502422-15502673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D2A
Entrez Gene ID
57545
Gene Name
coiled-coil and C2 domain containing 2A
Gene Aliases
JBTS9, MKS6
UniGene
Hs.590928
Gene Chromosome Location
Chr.4: 15468660-15601971 on Build GRCh38
Transcripts
NM_001080522.2 XM_005248177.1 XM_011513872.2 XM_017008482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5455277 c.160C>T p.R54C Chr.4: 15502488-15502488 on Build GRCh38
3128658 c.257C>T p.P86L Chr.4: 15502438-15502438 on Build GRCh38
6862353 c.152T>A p.M51K Chr.4: 15502480-15502480 on Build GRCh38
6862352 c.299T>A p.M100K Chr.4: 15502480-15502480 on Build GRCh38
3128659 c.110C>T p.P37L Chr.4: 15502438-15502438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034789313 Chr.4: 15502545-15502545 on Build GRCh38
rs745465296 Chr.4: 15502453-15502453 on Build GRCh38
rs761699527 Chr.4: 15502480-15502480 on Build GRCh38
rs566281691 Chr.4: 15502489-15502489 on Build GRCh38
rs367699902 Chr.4: 15502488-15502488 on Build GRCh38
rs906727368 Chr.4: 15502610-15502610 on Build GRCh38
rs999460059 Chr.4: 15502550-15502550 on Build GRCh38
rs765915288 Chr.4: 15502544-15502545 on Build GRCh38
rs556544210 Chr.4: 15502571-15502571 on Build GRCh38
rs753884581 Chr.4: 15502493-15502493 on Build GRCh38
rs764096982 Chr.4: 15502476-15502476 on Build GRCh38
rs376438052 Chr.4: 15502467-15502467 on Build GRCh38
rs751191380 Chr.4: 15502546-15502551 on Build GRCh38
rs201288603 Chr.4: 15502549-15502549 on Build GRCh38
rs201807781 Chr.4: 15502526-15502526 on Build GRCh38
rs142389729 Chr.4: 15502637-15502637 on Build GRCh38
rs987594766 Chr.4: 15502460-15502460 on Build GRCh38
rs370014549 Chr.4: 15502479-15502479 on Build GRCh38
rs137878385 Chr.4: 15502513-15502513 on Build GRCh38
rs373765548 Chr.4: 15502486-15502486 on Build GRCh38
rs576685639 Chr.4: 15502617-15502617 on Build GRCh38
rs748797062 Chr.4: 15502558-15502558 on Build GRCh38
rs768351928 Chr.4: 15502566-15502566 on Build GRCh38
rs755107436 Chr.4: 15502506-15502506 on Build GRCh38
rs182119485 Chr.4: 15502463-15502463 on Build GRCh38
rs886043867 Chr.4: 15502455-15502455 on Build GRCh38
rs757807971 Chr.4: 15502472-15502478 on Build GRCh38
rs200273579 Chr.4: 15502514-15502514 on Build GRCh38
rs886113667 Chr.4: 15502496-15502496 on Build GRCh38
rs911337809 Chr.4: 15502603-15502603 on Build GRCh38
rs779898482 Chr.4: 15502553-15502553 on Build GRCh38
rs368001493 Chr.4: 15502546-15502546 on Build GRCh38
rs1031382732 Chr.4: 15502551-15502551 on Build GRCh38
rs778148568 Chr.4: 15502534-15502534 on Build GRCh38
rs767278928 Chr.4: 15502484-15502484 on Build GRCh38
rs1012111099 Chr.4: 15502548-15502548 on Build GRCh38
rs762588216 Chr.4: 15502474-15502474 on Build GRCh38
rs955417539 Chr.4: 15502555-15502555 on Build GRCh38

View Full Product Details