Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 89350835-89351076 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP6
Entrez Gene ID
1848
Gene Name
dual specificity phosphatase 6
Gene Aliases
HH19, MKP3, PYST1
UniGene
Hs.298654
Gene Chromosome Location
Chr.12: 89347825-89352859 on Build GRCh38
Transcripts
NM_001946.3 NM_022652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4045283 c.556C>T p.R186* Chr.12: 89350870-89350870 on Build GRCh38
3987251 c.526T>C p.S176P Chr.12: 89350900-89350900 on Build GRCh38
1995008 c.523G>A p.D175N Chr.12: 89350903-89350903 on Build GRCh38
4969338 c.477G>C p.S159S Chr.12: 89350949-89350949 on Build GRCh38
4147504 c.550C>A p.L184I Chr.12: 89350876-89350876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751327509 Chr.12: 89350871-89350871 on Build GRCh38
rs772530383 Chr.12: 89351015-89351015 on Build GRCh38
rs561393208 Chr.12: 89350859-89350859 on Build GRCh38
rs149424762 Chr.12: 89350892-89350892 on Build GRCh38
rs808820 Chr.12: 89351049-89351049 on Build GRCh38
rs143567248 Chr.12: 89350863-89350863 on Build GRCh38
rs747182457 Chr.12: 89351036-89351036 on Build GRCh38
rs369073220 Chr.12: 89350949-89350949 on Build GRCh38
rs777206861 Chr.12: 89350981-89350981 on Build GRCh38
rs975617984 Chr.12: 89351017-89351017 on Build GRCh38
rs772620260 Chr.12: 89350920-89350920 on Build GRCh38
rs147303243 Chr.12: 89350861-89350861 on Build GRCh38
rs748751373 Chr.12: 89350989-89350989 on Build GRCh38
rs754716559 Chr.12: 89350956-89350956 on Build GRCh38
rs746212629 Chr.12: 89350907-89350907 on Build GRCh38
rs775828984 Chr.12: 89350931-89350931 on Build GRCh38
rs779272748 Chr.12: 89351003-89351003 on Build GRCh38
rs951013020 Chr.12: 89350967-89350967 on Build GRCh38
rs1019778679 Chr.12: 89351001-89351001 on Build GRCh38
rs144857723 Chr.12: 89350898-89350898 on Build GRCh38
rs767349460 Chr.12: 89350958-89350958 on Build GRCh38
rs780600285 Chr.12: 89351028-89351028 on Build GRCh38
rs965274509 Chr.12: 89351005-89351005 on Build GRCh38
rs766233542 Chr.12: 89350953-89350953 on Build GRCh38
rs375413401 Chr.12: 89351043-89351043 on Build GRCh38
rs776709235 Chr.12: 89351046-89351046 on Build GRCh38
rs770087 Chr.12: 89350996-89350996 on Build GRCh38
rs751583011 Chr.12: 89350955-89350955 on Build GRCh38
rs766152118 Chr.12: 89350870-89350870 on Build GRCh38
rs143946794 Chr.12: 89350860-89350860 on Build GRCh38
rs771353647 Chr.12: 89350901-89350901 on Build GRCh38
rs766734396 Chr.12: 89350919-89350920 on Build GRCh38
rs752316502 Chr.12: 89350965-89350965 on Build GRCh38
rs764233683 Chr.12: 89350934-89350934 on Build GRCh38
rs774709131 Chr.12: 89350905-89350905 on Build GRCh38
rs139318648 Chr.12: 89350881-89350881 on Build GRCh38
rs776876892 Chr.12: 89350943-89350943 on Build GRCh38
rs61754239 Chr.12: 89350918-89350918 on Build GRCh38
rs372462926 Chr.12: 89351004-89351004 on Build GRCh38
rs546367623 Chr.12: 89350888-89350888 on Build GRCh38
rs758302710 Chr.12: 89350867-89350867 on Build GRCh38
rs748460884 Chr.12: 89350896-89350896 on Build GRCh38
rs373643334 Chr.12: 89351035-89351035 on Build GRCh38

View Full Product Details