Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94765022-94765247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L4
Entrez Gene ID
286148
Gene Name
dpy-19 like 4 (C. elegans)
Gene Aliases
-
UniGene
Hs.567828
Gene Chromosome Location
Chr.8: 94719708-94793848 on Build GRCh38
Transcripts
NM_181787.2 XM_005250894.4 XM_005250895.3 XM_011516983.2 XM_011516984.2 XM_017013351.1 XM_017013352.1 XM_017013353.1 XM_017013354.1 XM_017013355.1 XM_017013356.1 XM_017013357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652006 c.904C>T p.L302F Chr.8: 94765216-94765216 on Build GRCh38
1102655 c.891C>A p.I297I Chr.8: 94765203-94765203 on Build GRCh38
6649089 c.898delT p.S301fs*21 Chr.8: 94765210-94765210 on Build GRCh38
3652005 c.903C>T p.S301S Chr.8: 94765215-94765215 on Build GRCh38
6055282 c.895A>T p.I299L Chr.8: 94765207-94765207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051268701 Chr.8: 94765064-94765064 on Build GRCh38
rs572654372 Chr.8: 94765135-94765135 on Build GRCh38
rs200567931 Chr.8: 94765176-94765176 on Build GRCh38
rs765069994 Chr.8: 94765132-94765132 on Build GRCh38
rs754689543 Chr.8: 94765155-94765155 on Build GRCh38
rs772434466 Chr.8: 94765188-94765188 on Build GRCh38
rs940185878 Chr.8: 94765081-94765081 on Build GRCh38
rs747057110 Chr.8: 94765207-94765207 on Build GRCh38
rs878920024 Chr.8: 94765073-94765073 on Build GRCh38
rs202225856 Chr.8: 94765174-94765174 on Build GRCh38
rs768790617 Chr.8: 94765216-94765216 on Build GRCh38
rs746426201 Chr.8: 94765187-94765187 on Build GRCh38
rs758231116 Chr.8: 94765139-94765139 on Build GRCh38
rs751018037 Chr.8: 94765152-94765152 on Build GRCh38
rs747736844 Chr.8: 94765163-94765163 on Build GRCh38
rs754116355 Chr.8: 94765160-94765164 on Build GRCh38
rs969245758 Chr.8: 94765062-94765062 on Build GRCh38
rs766580962 Chr.8: 94765158-94765161 on Build GRCh38
rs376788173 Chr.8: 94765158-94765158 on Build GRCh38
rs756188473 Chr.8: 94765150-94765151 on Build GRCh38
rs765957357 Chr.8: 94765151-94765151 on Build GRCh38
rs200039340 Chr.8: 94765196-94765196 on Build GRCh38
rs891334164 Chr.8: 94765068-94765068 on Build GRCh38

View Full Product Details