Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 65762672-65762921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH7
Entrez Gene ID
1005
Gene Name
cadherin 7
Gene Aliases
CDH7L1
UniGene
Hs.541003
Gene Chromosome Location
Chr.18: 65750252-65885136 on Build GRCh38
Transcripts
NM_001317214.1 NM_004361.3 NM_033646.2 XM_017025523.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4154003 c.42A>G p.L14L Chr.18: 65762884-65762884 on Build GRCh38
1630643 c.41T>C p.L14P Chr.18: 65762883-65762883 on Build GRCh38
6421413 c.56T>G p.L19R Chr.18: 65762898-65762898 on Build GRCh38
5700179 c.10G>A p.G4S Chr.18: 65762852-65762852 on Build GRCh38
3362578 c.28C>G p.H10D Chr.18: 65762870-65762870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894361486 Chr.18: 65762752-65762752 on Build GRCh38
rs753653033 Chr.18: 65762835-65762835 on Build GRCh38
rs757856072 Chr.18: 65762890-65762890 on Build GRCh38
rs752664317 Chr.18: 65762882-65762882 on Build GRCh38
rs1012952981 Chr.18: 65762753-65762753 on Build GRCh38
rs369635030 Chr.18: 65762800-65762800 on Build GRCh38
rs767791804 Chr.18: 65762780-65762780 on Build GRCh38
rs61741929 Chr.18: 65762893-65762893 on Build GRCh38
rs147058533 Chr.18: 65762820-65762820 on Build GRCh38
rs766577048 Chr.18: 65762832-65762832 on Build GRCh38
rs938634582 Chr.18: 65762735-65762735 on Build GRCh38
rs895896644 Chr.18: 65762740-65762740 on Build GRCh38
rs904287653 Chr.18: 65762777-65762777 on Build GRCh38
rs1032299494 Chr.18: 65762839-65762839 on Build GRCh38
rs141768254 Chr.18: 65762798-65762798 on Build GRCh38
rs114985920 Chr.18: 65762848-65762848 on Build GRCh38
rs199553609 Chr.18: 65762886-65762886 on Build GRCh38
rs904501512 Chr.18: 65762786-65762786 on Build GRCh38
rs1031019491 Chr.18: 65762888-65762888 on Build GRCh38
rs997832923 Chr.18: 65762871-65762871 on Build GRCh38
rs1012720997 Chr.18: 65762765-65762765 on Build GRCh38
rs761205882 Chr.18: 65762832-65762835 on Build GRCh38
rs750577091 Chr.18: 65762819-65762819 on Build GRCh38
rs767724012 Chr.18: 65762811-65762811 on Build GRCh38
rs200963885 Chr.18: 65762831-65762832 on Build GRCh38
rs569702559 Chr.18: 65762804-65762804 on Build GRCh38
rs867299931 Chr.18: 65762845-65762845 on Build GRCh38
rs1057277196 Chr.18: 65762730-65762730 on Build GRCh38
rs757353839 Chr.18: 65762776-65762776 on Build GRCh38
rs998553717 Chr.18: 65762787-65762787 on Build GRCh38
rs767813460 Chr.18: 65762832-65762832 on Build GRCh38
rs766765265 Chr.18: 65762834-65762835 on Build GRCh38
rs929084989 Chr.18: 65762723-65762723 on Build GRCh38
rs1000458853 Chr.18: 65762817-65762817 on Build GRCh38
rs754425783 Chr.18: 65762841-65762842 on Build GRCh38

View Full Product Details