Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 31262952-31263197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCDC1
Entrez Gene ID
341019
Gene Name
doublecortin domain containing 1
Gene Aliases
-
UniGene
Hs.127405
Gene Chromosome Location
Chr.11: 31262624-31369810 on Build GRCh38
Transcripts
NM_181807.3

SNP Detail

SNP ID Chromosome Location
rs752911745 Chr.11: 31263108-31263108 on Build GRCh38
rs912463722 Chr.11: 31263148-31263148 on Build GRCh38
rs769412097 Chr.11: 31263067-31263067 on Build GRCh38
rs534456844 Chr.11: 31263025-31263025 on Build GRCh38
rs181878016 Chr.11: 31263048-31263048 on Build GRCh38
rs774803528 Chr.11: 31263099-31263099 on Build GRCh38
rs141209313 Chr.11: 31263072-31263072 on Build GRCh38
rs200196165 Chr.11: 31263112-31263112 on Build GRCh38
rs936335215 Chr.11: 31263161-31263161 on Build GRCh38
rs767587840 Chr.11: 31263012-31263012 on Build GRCh38
rs368936486 Chr.11: 31263011-31263011 on Build GRCh38
rs199802073 Chr.11: 31263076-31263076 on Build GRCh38
rs975739086 Chr.11: 31263153-31263153 on Build GRCh38
rs758167639 Chr.11: 31263046-31263046 on Build GRCh38
rs150306954 Chr.11: 31263136-31263136 on Build GRCh38
rs960222909 Chr.11: 31263037-31263037 on Build GRCh38
rs750337662 Chr.11: 31263041-31263041 on Build GRCh38
rs759763463 Chr.11: 31263102-31263102 on Build GRCh38
rs375627695 Chr.11: 31263098-31263098 on Build GRCh38
rs966357245 Chr.11: 31263056-31263056 on Build GRCh38
rs920906461 Chr.11: 31263154-31263154 on Build GRCh38
rs369329098 Chr.11: 31263054-31263054 on Build GRCh38
rs767787537 Chr.11: 31263106-31263106 on Build GRCh38
rs1009762042 Chr.11: 31262999-31262999 on Build GRCh38
rs561215227 Chr.11: 31263149-31263149 on Build GRCh38
rs780690958 Chr.11: 31262998-31262998 on Build GRCh38
rs1053541119 Chr.11: 31262981-31262981 on Build GRCh38
rs186244171 Chr.11: 31263146-31263146 on Build GRCh38
rs772736810 Chr.11: 31263064-31263065 on Build GRCh38
rs749914659 Chr.11: 31263002-31263002 on Build GRCh38
rs756120926 Chr.11: 31263019-31263019 on Build GRCh38
rs752789070 Chr.11: 31263014-31263014 on Build GRCh38
rs375989757 Chr.11: 31263023-31263023 on Build GRCh38
rs746731854 Chr.11: 31263053-31263053 on Build GRCh38
rs906817653 Chr.11: 31263003-31263003 on Build GRCh38
rs748254385 Chr.11: 31263044-31263044 on Build GRCh38

View Full Product Details