Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56854811-56855078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8A
Entrez Gene ID
731
Gene Name
complement component 8 alpha subunit
Gene Aliases
-
UniGene
Hs.93210
Gene Chromosome Location
Chr.1: 56854770-56918221 on Build GRCh38
Transcripts
NM_000562.2 XM_011542079.2 XM_017002234.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386253 c.11T>C p.V4A Chr.1: 56854912-56854912 on Build GRCh38
3491172 c.19T>A p.F7I Chr.1: 56854920-56854920 on Build GRCh38
309777 c.24C>G p.I8M Chr.1: 56854925-56854925 on Build GRCh38
911092 c.24C>A p.I8I Chr.1: 56854925-56854925 on Build GRCh38
4144108 c.57T>C p.T19T Chr.1: 56854958-56854958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759920430 Chr.1: 56855013-56855013 on Build GRCh38
rs1052034243 Chr.1: 56854976-56854976 on Build GRCh38
rs761933641 Chr.1: 56854855-56854855 on Build GRCh38
rs774336945 Chr.1: 56854851-56854851 on Build GRCh38
rs766792632 Chr.1: 56854879-56854879 on Build GRCh38
rs761125557 Chr.1: 56854875-56854875 on Build GRCh38
rs773564988 Chr.1: 56854963-56854965 on Build GRCh38
rs750962980 Chr.1: 56854872-56854872 on Build GRCh38
rs566517154 Chr.1: 56855042-56855042 on Build GRCh38
rs756722484 Chr.1: 56855026-56855026 on Build GRCh38
rs188012019 Chr.1: 56855049-56855049 on Build GRCh38
rs923948777 Chr.1: 56854841-56854841 on Build GRCh38
rs745734571 Chr.1: 56854936-56854936 on Build GRCh38
rs763708708 Chr.1: 56855017-56855017 on Build GRCh38
rs752419487 Chr.1: 56854883-56854883 on Build GRCh38
rs267598665 Chr.1: 56854952-56854952 on Build GRCh38
rs774621470 Chr.1: 56854961-56854961 on Build GRCh38
rs769603846 Chr.1: 56854942-56854942 on Build GRCh38
rs546389515 Chr.1: 56855019-56855019 on Build GRCh38
rs373550196 Chr.1: 56855012-56855012 on Build GRCh38
rs750395071 Chr.1: 56855030-56855030 on Build GRCh38
rs768812752 Chr.1: 56854953-56854953 on Build GRCh38
rs552319150 Chr.1: 56854885-56854885 on Build GRCh38
rs184240619 Chr.1: 56854987-56854987 on Build GRCh38
rs112742526 Chr.1: 56854981-56854981 on Build GRCh38
rs113852423 Chr.1: 56854980-56854980 on Build GRCh38
rs187731564 Chr.1: 56854862-56854862 on Build GRCh38
rs1036578051 Chr.1: 56854956-56854956 on Build GRCh38
rs143035893 Chr.1: 56854884-56854884 on Build GRCh38
rs761215078 Chr.1: 56854996-56854996 on Build GRCh38
rs993868280 Chr.1: 56854988-56854988 on Build GRCh38
rs772124654 Chr.1: 56854992-56854992 on Build GRCh38
rs369518355 Chr.1: 56854994-56854994 on Build GRCh38
rs192696675 Chr.1: 56854927-56854927 on Build GRCh38
rs779787811 Chr.1: 56854951-56854951 on Build GRCh38
rs780618145 Chr.1: 56855027-56855027 on Build GRCh38
rs887915389 Chr.1: 56854909-56854909 on Build GRCh38

View Full Product Details