Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12720732-12721003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL3
Entrez Gene ID
126767
Gene Name
arylacetamide deacetylase like 3
Gene Aliases
-
UniGene
Hs.464705
Gene Chromosome Location
Chr.1: 12716115-12728759 on Build GRCh38
Transcripts
NM_001103170.2 NR_111984.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6533576 c.28C>G p.R10G Chr.1: 12720906-12720906 on Build GRCh38
5502153 c.40G>T p.E14* Chr.1: 12720918-12720918 on Build GRCh38
7353373 c.5-2A>G p.? Chr.1: 12720881-12720881 on Build GRCh38
4565765 c.39_40GG>AA p.E14K Chr.1: 12720917-12720918 on Build GRCh38
5649893 c.51C>A p.S17S Chr.1: 12720929-12720929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750676977 Chr.1: 12720936-12720936 on Build GRCh38
rs952446290 Chr.1: 12720822-12720822 on Build GRCh38
rs549849868 Chr.1: 12720785-12720785 on Build GRCh38
rs778846928 Chr.1: 12720966-12720966 on Build GRCh38
rs775970102 Chr.1: 12720906-12720906 on Build GRCh38
rs753644401 Chr.1: 12720932-12720932 on Build GRCh38
rs552441177 Chr.1: 12720874-12720874 on Build GRCh38
rs756346985 Chr.1: 12720940-12720940 on Build GRCh38
rs746043903 Chr.1: 12720843-12720843 on Build GRCh38
rs748063569 Chr.1: 12720967-12720967 on Build GRCh38
rs768560484 Chr.1: 12720885-12720885 on Build GRCh38
rs755865972 Chr.1: 12720869-12720869 on Build GRCh38
rs377378795 Chr.1: 12720764-12720764 on Build GRCh38
rs767566887 Chr.1: 12720932-12720932 on Build GRCh38
rs771564598 Chr.1: 12720895-12720895 on Build GRCh38
rs375991061 Chr.1: 12720927-12720927 on Build GRCh38
rs374862028 Chr.1: 12720970-12720970 on Build GRCh38
rs935187588 Chr.1: 12720948-12720948 on Build GRCh38
rs907856896 Chr.1: 12720974-12720974 on Build GRCh38
rs990501194 Chr.1: 12720828-12720828 on Build GRCh38
rs781290432 Chr.1: 12720837-12720837 on Build GRCh38
rs367820626 Chr.1: 12720814-12720814 on Build GRCh38
rs1036978799 Chr.1: 12720818-12720818 on Build GRCh38
rs949126651 Chr.1: 12720901-12720901 on Build GRCh38
rs772756914 Chr.1: 12720897-12720897 on Build GRCh38
rs200734569 Chr.1: 12720904-12720904 on Build GRCh38
rs199614160 Chr.1: 12720954-12720954 on Build GRCh38
rs747645917 Chr.1: 12720893-12720893 on Build GRCh38
rs547389951 Chr.1: 12720929-12720929 on Build GRCh38
rs764726299 Chr.1: 12720913-12720913 on Build GRCh38
rs567249050 Chr.1: 12720930-12720930 on Build GRCh38
rs544009869 Chr.1: 12720790-12720790 on Build GRCh38
rs916291170 Chr.1: 12720871-12720871 on Build GRCh38
rs1029576839 Chr.1: 12720955-12720956 on Build GRCh38
rs779942887 Chr.1: 12720894-12720894 on Build GRCh38
rs1050464004 Chr.1: 12720972-12720972 on Build GRCh38
rs907882862 Chr.1: 12720791-12720791 on Build GRCh38
rs547636769 Chr.1: 12720784-12720784 on Build GRCh38
rs575046072 Chr.1: 12720781-12720781 on Build GRCh38
rs369765539 Chr.1: 12720951-12720951 on Build GRCh38
rs760312708 Chr.1: 12720902-12720902 on Build GRCh38
rs773892308 Chr.1: 12720891-12720891 on Build GRCh38
rs563583119 Chr.1: 12720788-12720788 on Build GRCh38
rs140815157 Chr.1: 12720907-12720907 on Build GRCh38
rs867622256 Chr.1: 12720963-12720963 on Build GRCh38
rs756066132 Chr.1: 12720843-12720843 on Build GRCh38
rs754729973 Chr.1: 12720964-12720964 on Build GRCh38

View Full Product Details