Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24038309-24038554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP1B
Entrez Gene ID
2281
Gene Name
FK506 binding protein 1B
Gene Aliases
FKBP12.6, FKBP1L, OTK4, PKBP1L, PPIase
UniGene
Hs.709461
Gene Chromosome Location
Chr.2: 24033205-24067743 on Build GRCh38
Transcripts
XM_017003594.1 NM_001142319.1 NM_025203.2 XM_017005029.1

Gene Symbol
WDCP
Entrez Gene ID
80304
Gene Name
WD repeat and coiled coil containing
Gene Aliases
C2orf44, PP384
UniGene
Hs.24624
Gene Chromosome Location
Chr.2: 24029336-24047426 on Build GRCh38
Transcripts
XM_017003594.1 NM_001142319.1 NM_025203.2 XM_017005029.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1406817 c.1081T>A p.C361S Chr.2: 24038414-24038414 on Build GRCh38
5673645 c.1139T>G p.I380S Chr.2: 24038356-24038356 on Build GRCh38
5601151 c.1111C>T p.P371S Chr.2: 24038384-24038384 on Build GRCh38
4519892 c.1032G>A p.L344L Chr.2: 24038463-24038463 on Build GRCh38
2910839 c.1089A>G p.I363M Chr.2: 24038406-24038406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755870703 Chr.2: 24038422-24038422 on Build GRCh38
rs762190972 Chr.2: 24038394-24038394 on Build GRCh38
rs765584189 Chr.2: 24038507-24038507 on Build GRCh38
rs371021807 Chr.2: 24038443-24038443 on Build GRCh38
rs779527289 Chr.2: 24038407-24038407 on Build GRCh38
rs777585038 Chr.2: 24038430-24038430 on Build GRCh38
rs532108271 Chr.2: 24038406-24038406 on Build GRCh38
rs373591251 Chr.2: 24038353-24038353 on Build GRCh38
rs145288775 Chr.2: 24038521-24038521 on Build GRCh38
rs754230399 Chr.2: 24038387-24038387 on Build GRCh38
rs1044583763 Chr.2: 24038520-24038520 on Build GRCh38
rs140791647 Chr.2: 24038523-24038523 on Build GRCh38
rs747925134 Chr.2: 24038413-24038413 on Build GRCh38
rs140402918 Chr.2: 24038484-24038484 on Build GRCh38
rs201664806 Chr.2: 24038517-24038517 on Build GRCh38
rs749231297 Chr.2: 24038437-24038437 on Build GRCh38
rs564999222 Chr.2: 24038400-24038400 on Build GRCh38
rs776875471 Chr.2: 24038502-24038502 on Build GRCh38
rs376816524 Chr.2: 24038410-24038410 on Build GRCh38
rs780771485 Chr.2: 24038412-24038412 on Build GRCh38
rs201871067 Chr.2: 24038503-24038503 on Build GRCh38
rs751281338 Chr.2: 24038408-24038408 on Build GRCh38
rs769024083 Chr.2: 24038500-24038500 on Build GRCh38
rs764589613 Chr.2: 24038361-24038361 on Build GRCh38
rs772341651 Chr.2: 24038438-24038438 on Build GRCh38
rs1037017289 Chr.2: 24038416-24038416 on Build GRCh38

View Full Product Details