Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78885497-78885736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSB
Entrez Gene ID
411
Gene Name
arylsulfatase B
Gene Aliases
ASB, G4S, MPS6
UniGene
Hs.149103
Gene Chromosome Location
Chr.5: 78777209-78986534 on Build GRCh38
Transcripts
NM_000046.3 NM_198709.2 XM_011543390.1 XM_011543391.2 XM_011543392.2 XM_011543393.1 XM_017009471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4778466 c.1107G>T p.K369N Chr.5: 78885619-78885619 on Build GRCh38
1634327 c.1107G>T p.K369N Chr.5: 78885619-78885619 on Build GRCh38
1620569 c.1042A>T p.I348F Chr.5: 78885684-78885684 on Build GRCh38
1486988 c.1059G>T p.W353C Chr.5: 78885667-78885667 on Build GRCh38
5555331 c.1021G>C p.G341R Chr.5: 78885705-78885705 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371916604 Chr.5: 78885573-78885573 on Build GRCh38
rs16876029 Chr.5: 78885658-78885658 on Build GRCh38
rs772218687 Chr.5: 78885613-78885613 on Build GRCh38
rs1071598 Chr.5: 78885600-78885600 on Build GRCh38
rs752366493 Chr.5: 78885557-78885557 on Build GRCh38
rs200040980 Chr.5: 78885603-78885603 on Build GRCh38
rs756793742 Chr.5: 78885575-78885575 on Build GRCh38
rs756671977 Chr.5: 78885674-78885674 on Build GRCh38
rs148067062 Chr.5: 78885692-78885692 on Build GRCh38
rs765177876 Chr.5: 78885539-78885539 on Build GRCh38
rs780432111 Chr.5: 78885601-78885601 on Build GRCh38
rs765119051 Chr.5: 78885643-78885643 on Build GRCh38
rs768620820 Chr.5: 78885529-78885529 on Build GRCh38
rs749593643 Chr.5: 78885702-78885702 on Build GRCh38
rs746396210 Chr.5: 78885583-78885583 on Build GRCh38
rs368078480 Chr.5: 78885691-78885691 on Build GRCh38
rs1065757 Chr.5: 78885654-78885654 on Build GRCh38
rs145433056 Chr.5: 78885679-78885679 on Build GRCh38
rs144879531 Chr.5: 78885640-78885640 on Build GRCh38
rs759360225 Chr.5: 78885537-78885537 on Build GRCh38
rs774644004 Chr.5: 78885606-78885606 on Build GRCh38
rs993937950 Chr.5: 78885584-78885584 on Build GRCh38
rs149886659 Chr.5: 78885604-78885604 on Build GRCh38
rs766168716 Chr.5: 78885534-78885534 on Build GRCh38
rs1071598 Chr.5: 78885600-78885600 on Build GRCh38
rs752599167 Chr.5: 78885645-78885645 on Build GRCh38
rs753861380 Chr.5: 78885536-78885536 on Build GRCh38
rs79970603 Chr.5: 78885678-78885678 on Build GRCh38
rs780380678 Chr.5: 78885693-78885693 on Build GRCh38
rs750977557 Chr.5: 78885574-78885574 on Build GRCh38
rs758055045 Chr.5: 78885566-78885566 on Build GRCh38
rs776690571 Chr.5: 78885630-78885630 on Build GRCh38
rs781510986 Chr.5: 78885582-78885582 on Build GRCh38
rs769067669 Chr.5: 78885703-78885703 on Build GRCh38
rs140923667 Chr.5: 78885648-78885648 on Build GRCh38

View Full Product Details