Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82389857-82390094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
METTL25
Entrez Gene ID
84190
Gene Name
methyltransferase like 25
Gene Aliases
C12orf26
UniGene
Hs.506222
Gene Chromosome Location
Chr.12: 82358497-82479239 on Build GRCh38
Transcripts
NM_001319675.1 NM_032230.2 XM_005269186.4 XM_011538827.2 XM_011538828.2 XM_011538829.2 XM_017020020.1 XM_017020021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1993789 c.528G>A p.K176K Chr.12: 82389919-82389919 on Build GRCh38
1993788 c.508G>A p.A170T Chr.12: 82389899-82389899 on Build GRCh38
3813113 c.490G>C p.E164Q Chr.12: 82389881-82389881 on Build GRCh38
943957 c.467C>T p.S156F Chr.12: 82389858-82389858 on Build GRCh38
7154252 c.531+1G>A p.? Chr.12: 82389923-82389923 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777951656 Chr.12: 82389909-82389909 on Build GRCh38
rs946993297 Chr.12: 82390041-82390041 on Build GRCh38
rs760756432 Chr.12: 82389942-82389942 on Build GRCh38
rs756516559 Chr.12: 82389906-82389906 on Build GRCh38
rs375677021 Chr.12: 82389964-82389964 on Build GRCh38
rs747690335 Chr.12: 82389903-82389903 on Build GRCh38
rs199539756 Chr.12: 82389934-82389934 on Build GRCh38
rs765108028 Chr.12: 82389949-82389949 on Build GRCh38
rs557748288 Chr.12: 82389936-82389936 on Build GRCh38
rs781060214 Chr.12: 82389901-82389901 on Build GRCh38
rs775791284 Chr.12: 82389940-82389940 on Build GRCh38
rs369465475 Chr.12: 82389884-82389884 on Build GRCh38
rs1040676479 Chr.12: 82389892-82389892 on Build GRCh38
rs374311937 Chr.12: 82389960-82389960 on Build GRCh38
rs10778908 Chr.12: 82389889-82389889 on Build GRCh38
rs902665391 Chr.12: 82390009-82390009 on Build GRCh38
rs749800202 Chr.12: 82389937-82389937 on Build GRCh38
rs766486092 Chr.12: 82389965-82389965 on Build GRCh38
rs771477466 Chr.12: 82389923-82389923 on Build GRCh38
rs749832501 Chr.12: 82389922-82389922 on Build GRCh38
rs922665068 Chr.12: 82389902-82389902 on Build GRCh38
rs987830006 Chr.12: 82389995-82389995 on Build GRCh38
rs915482791 Chr.12: 82390008-82390008 on Build GRCh38
rs188729912 Chr.12: 82389974-82389974 on Build GRCh38
rs1003840351 Chr.12: 82390026-82390026 on Build GRCh38
rs373003458 Chr.12: 82389891-82389891 on Build GRCh38
rs761775867 Chr.12: 82389913-82389913 on Build GRCh38

View Full Product Details