Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111883676-111883949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf1
Entrez Gene ID
64776
Gene Name
chromosome 11 open reading frame 1
Gene Aliases
-
UniGene
Hs.17546
Gene Chromosome Location
Chr.11: 111879190-111884083 on Build GRCh38
Transcripts
NM_022761.2 XM_011542957.2 XM_011542958.2 XM_017018129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4666586 c.352C>T p.R118* Chr.11: 111883779-111883779 on Build GRCh38
3791185 c.355G>A p.E119K Chr.11: 111883782-111883782 on Build GRCh38
3998231 c.423A>C p.S141S Chr.11: 111883850-111883850 on Build GRCh38
6613972 c.348T>G p.F116L Chr.11: 111883775-111883775 on Build GRCh38
5893532 c.398C>T p.P133L Chr.11: 111883825-111883825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782175415 Chr.11: 111883886-111883886 on Build GRCh38
rs782793057 Chr.11: 111883801-111883801 on Build GRCh38
rs782501825 Chr.11: 111883752-111883752 on Build GRCh38
rs996989141 Chr.11: 111883751-111883751 on Build GRCh38
rs782014780 Chr.11: 111883894-111883894 on Build GRCh38
rs782258472 Chr.11: 111883857-111883857 on Build GRCh38
rs550765828 Chr.11: 111883714-111883714 on Build GRCh38
rs969267615 Chr.11: 111883876-111883876 on Build GRCh38
rs781840394 Chr.11: 111883824-111883824 on Build GRCh38
rs782369504 Chr.11: 111883780-111883780 on Build GRCh38
rs142750898 Chr.11: 111883862-111883862 on Build GRCh38
rs370959868 Chr.11: 111883841-111883841 on Build GRCh38
rs782736350 Chr.11: 111883828-111883828 on Build GRCh38
rs1044372186 Chr.11: 111883733-111883733 on Build GRCh38
rs376314856 Chr.11: 111883904-111883904 on Build GRCh38
rs1045282 Chr.11: 111883850-111883850 on Build GRCh38
rs782638393 Chr.11: 111883749-111883749 on Build GRCh38
rs145116726 Chr.11: 111883770-111883770 on Build GRCh38
rs548531796 Chr.11: 111883869-111883869 on Build GRCh38
rs782807714 Chr.11: 111883751-111883752 on Build GRCh38
rs370385310 Chr.11: 111883888-111883888 on Build GRCh38
rs782295307 Chr.11: 111883897-111883897 on Build GRCh38
rs373898544 Chr.11: 111883901-111883901 on Build GRCh38
rs782534998 Chr.11: 111883748-111883748 on Build GRCh38
rs142305825 Chr.11: 111883793-111883793 on Build GRCh38
rs782071949 Chr.11: 111883786-111883786 on Build GRCh38
rs782095154 Chr.11: 111883730-111883730 on Build GRCh38
rs200474782 Chr.11: 111883803-111883803 on Build GRCh38
rs782259753 Chr.11: 111883717-111883717 on Build GRCh38
rs781873261 Chr.11: 111883752-111883752 on Build GRCh38
rs782138458 Chr.11: 111883812-111883812 on Build GRCh38
rs782339956 Chr.11: 111883768-111883768 on Build GRCh38
rs200985273 Chr.11: 111883796-111883796 on Build GRCh38
rs782100893 Chr.11: 111883906-111883906 on Build GRCh38
rs145926656 Chr.11: 111883827-111883827 on Build GRCh38
rs782356924 Chr.11: 111883787-111883787 on Build GRCh38
rs568752916 Chr.11: 111883911-111883911 on Build GRCh38
rs782729981 Chr.11: 111883818-111883818 on Build GRCh38
rs782630637 Chr.11: 111883852-111883852 on Build GRCh38
rs781994736 Chr.11: 111883782-111883782 on Build GRCh38
rs782602393 Chr.11: 111883885-111883885 on Build GRCh38
rs782291087 Chr.11: 111883779-111883779 on Build GRCh38
rs782294085 Chr.11: 111883839-111883839 on Build GRCh38
rs193069595 Chr.11: 111883761-111883761 on Build GRCh38
rs782483556 Chr.11: 111883756-111883756 on Build GRCh38
rs782699056 Chr.11: 111883731-111883731 on Build GRCh38
rs781807626 Chr.11: 111883735-111883735 on Build GRCh38

View Full Product Details