Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130568359-130568618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A6
Entrez Gene ID
131873
Gene Name
collagen type VI alpha 6
Gene Aliases
-
UniGene
Hs.591282
Gene Chromosome Location
Chr.3: 130516709-130678135 on Build GRCh38
Transcripts
NM_001102608.1 XM_005247121.4 XM_011512423.2 XM_011512424.2 XM_011512425.2 XM_011512426.2 XM_011512428.2 XM_011512431.2 XM_011512432.2 XM_017005711.1 XM_017005712.1 XM_017005713.1 XM_017005714.1 XM_017005715.1 XM_017005716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4839733 c.2192A>G p.D731G Chr.3: 130568395-130568395 on Build GRCh38
1418960 c.2275G>A p.G759S Chr.3: 130568478-130568478 on Build GRCh38
1418958 c.2261C>A p.S754Y Chr.3: 130568464-130568464 on Build GRCh38
274627 c.2176C>A p.L726I Chr.3: 130568379-130568379 on Build GRCh38
4624965 c.2342T>C p.F781S Chr.3: 130568545-130568545 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566520876 Chr.3: 130568392-130568392 on Build GRCh38
rs995128581 Chr.3: 130568428-130568428 on Build GRCh38
rs370031148 Chr.3: 130568393-130568393 on Build GRCh38
rs758083788 Chr.3: 130568473-130568473 on Build GRCh38
rs747940604 Chr.3: 130568382-130568382 on Build GRCh38
rs776862405 Chr.3: 130568562-130568562 on Build GRCh38
rs780740343 Chr.3: 130568507-130568507 on Build GRCh38
rs750757392 Chr.3: 130568570-130568570 on Build GRCh38
rs763521970 Chr.3: 130568412-130568412 on Build GRCh38
rs765204569 Chr.3: 130568465-130568465 on Build GRCh38
rs764240714 Chr.3: 130568526-130568527 on Build GRCh38
rs756932668 Chr.3: 130568495-130568495 on Build GRCh38
rs759362791 Chr.3: 130568491-130568491 on Build GRCh38
rs773593204 Chr.3: 130568546-130568546 on Build GRCh38
rs555485052 Chr.3: 130568459-130568459 on Build GRCh38
rs763866351 Chr.3: 130568597-130568597 on Build GRCh38
rs966503329 Chr.3: 130568524-130568524 on Build GRCh38
rs762099335 Chr.3: 130568415-130568415 on Build GRCh38
rs748534224 Chr.3: 130568531-130568531 on Build GRCh38
rs767812525 Chr.3: 130568429-130568429 on Build GRCh38
rs556475574 Chr.3: 130568559-130568559 on Build GRCh38
rs778915716 Chr.3: 130568381-130568381 on Build GRCh38
rs776178739 Chr.3: 130568407-130568407 on Build GRCh38
rs1022318956 Chr.3: 130568489-130568489 on Build GRCh38
rs752734162 Chr.3: 130568592-130568592 on Build GRCh38
rs746683005 Chr.3: 130568491-130568491 on Build GRCh38
rs201087058 Chr.3: 130568595-130568595 on Build GRCh38
rs373377669 Chr.3: 130568599-130568599 on Build GRCh38
rs200242417 Chr.3: 130568430-130568430 on Build GRCh38
rs201380592 Chr.3: 130568535-130568535 on Build GRCh38
rs777221597 Chr.3: 130568385-130568385 on Build GRCh38
rs760929088 Chr.3: 130568439-130568439 on Build GRCh38
rs774713000 Chr.3: 130568414-130568414 on Build GRCh38
rs921699418 Chr.3: 130568598-130568598 on Build GRCh38
rs768928035 Chr.3: 130568502-130568502 on Build GRCh38
rs374095055 Chr.3: 130568551-130568551 on Build GRCh38
rs776849020 Chr.3: 130568565-130568567 on Build GRCh38
rs373883247 Chr.3: 130568449-130568449 on Build GRCh38
rs371129727 Chr.3: 130568440-130568440 on Build GRCh38
rs752541892 Chr.3: 130568472-130568472 on Build GRCh38
rs771765065 Chr.3: 130568384-130568384 on Build GRCh38
rs774081404 Chr.3: 130568452-130568453 on Build GRCh38
rs755742964 Chr.3: 130568485-130568485 on Build GRCh38
rs376333031 Chr.3: 130568504-130568504 on Build GRCh38
rs61730502 Chr.3: 130568519-130568519 on Build GRCh38
rs769140737 Chr.3: 130568413-130568413 on Build GRCh38
rs759620011 Chr.3: 130568563-130568563 on Build GRCh38
rs573547736 Chr.3: 130568516-130568516 on Build GRCh38
rs978007456 Chr.3: 130568558-130568558 on Build GRCh38
rs974941265 Chr.3: 130568526-130568526 on Build GRCh38
rs765701051 Chr.3: 130568560-130568560 on Build GRCh38
rs774800733 Chr.3: 130568520-130568520 on Build GRCh38

View Full Product Details