Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137880375-137880646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L2
Entrez Gene ID
64764
Gene Name
cAMP responsive element binding protein 3 like 2
Gene Aliases
BBF2H7
UniGene
Hs.490273
Gene Chromosome Location
Chr.7: 137874979-138002101 on Build GRCh38
Transcripts
NM_001318246.1 NM_194071.3 XM_005250541.4 XM_017012540.1 XM_017012541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4420880 c.1541G>A p.R514K Chr.7: 137880498-137880498 on Build GRCh38
2859686 c.1495G>A p.A499T Chr.7: 137880544-137880544 on Build GRCh38
2859685 c.1536C>T p.L512L Chr.7: 137880503-137880503 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768524282 Chr.7: 137880554-137880554 on Build GRCh38
rs772307261 Chr.7: 137880434-137880434 on Build GRCh38
rs747151734 Chr.7: 137880544-137880544 on Build GRCh38
rs369276139 Chr.7: 137880549-137880549 on Build GRCh38
rs897570858 Chr.7: 137880457-137880457 on Build GRCh38
rs748709574 Chr.7: 137880499-137880499 on Build GRCh38
rs767040443 Chr.7: 137880460-137880460 on Build GRCh38
rs776710093 Chr.7: 137880439-137880439 on Build GRCh38
rs759090540 Chr.7: 137880596-137880596 on Build GRCh38
rs374387990 Chr.7: 137880495-137880495 on Build GRCh38
rs543766767 Chr.7: 137880461-137880461 on Build GRCh38
rs771117390 Chr.7: 137880545-137880545 on Build GRCh38
rs371909088 Chr.7: 137880436-137880436 on Build GRCh38
rs760990511 Chr.7: 137880437-137880437 on Build GRCh38
rs767379871 Chr.7: 137880579-137880579 on Build GRCh38
rs753419692 Chr.7: 137880479-137880479 on Build GRCh38
rs954015707 Chr.7: 137880601-137880601 on Build GRCh38
rs778003332 Chr.7: 137880542-137880542 on Build GRCh38
rs746043244 Chr.7: 137880552-137880552 on Build GRCh38
rs191006690 Chr.7: 137880575-137880575 on Build GRCh38
rs905374450 Chr.7: 137880517-137880517 on Build GRCh38
rs576726956 Chr.7: 137880602-137880602 on Build GRCh38
rs770638470 Chr.7: 137880463-137880463 on Build GRCh38
rs1036069110 Chr.7: 137880400-137880400 on Build GRCh38
rs773133391 Chr.7: 137880580-137880580 on Build GRCh38
rs972935806 Chr.7: 137880625-137880625 on Build GRCh38
rs375770659 Chr.7: 137880576-137880576 on Build GRCh38
rs766876279 Chr.7: 137880477-137880477 on Build GRCh38
rs202085147 Chr.7: 137880537-137880537 on Build GRCh38
rs1049676012 Chr.7: 137880484-137880484 on Build GRCh38
rs771211415 Chr.7: 137880438-137880438 on Build GRCh38
rs574770108 Chr.7: 137880459-137880459 on Build GRCh38
rs760236840 Chr.7: 137880585-137880585 on Build GRCh38
rs748482110 Chr.7: 137880454-137880454 on Build GRCh38
rs369590106 Chr.7: 137880612-137880612 on Build GRCh38
rs961891557 Chr.7: 137880623-137880623 on Build GRCh38
rs754536469 Chr.7: 137880603-137880603 on Build GRCh38
rs748583702 Chr.7: 137880430-137880430 on Build GRCh38
rs762859900 Chr.7: 137880452-137880452 on Build GRCh38
rs779686427 Chr.7: 137880456-137880457 on Build GRCh38
rs755421445 Chr.7: 137880491-137880491 on Build GRCh38
rs896782003 Chr.7: 137880617-137880617 on Build GRCh38
rs141738028 Chr.7: 137880458-137880458 on Build GRCh38
rs987085573 Chr.7: 137880608-137880608 on Build GRCh38

View Full Product Details