Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 64553223-64553462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFTPH
Entrez Gene ID
54812
Gene Name
aftiphilin
Gene Aliases
Nbla10388
UniGene
Hs.655167
Gene Chromosome Location
Chr.2: 64524305-64593005 on Build GRCh38
Transcripts
NM_001002243.2 NM_017657.4 NM_203437.3 XM_005264380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1592293 c.1795C>T p.R599* Chr.2: 64553269-64553269 on Build GRCh38
5730300 c.1862_1863insC p.S622fs*13 Chr.2: 64553336-64553337 on Build GRCh38
6261131 c.1828G>T p.E610* Chr.2: 64553302-64553302 on Build GRCh38
6266219 c.1921G>T p.A641S Chr.2: 64553395-64553395 on Build GRCh38
1149194 c.1759G>C p.A587P Chr.2: 64553233-64553233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764819647 Chr.2: 64553333-64553333 on Build GRCh38
rs757903472 Chr.2: 64553367-64553367 on Build GRCh38
rs753154038 Chr.2: 64553253-64553253 on Build GRCh38
rs747930756 Chr.2: 64553297-64553297 on Build GRCh38
rs746934237 Chr.2: 64553312-64553312 on Build GRCh38
rs375454324 Chr.2: 64553255-64553255 on Build GRCh38
rs185828021 Chr.2: 64553398-64553398 on Build GRCh38
rs989958765 Chr.2: 64553288-64553288 on Build GRCh38
rs778831869 Chr.2: 64553289-64553289 on Build GRCh38
rs371892827 Chr.2: 64553307-64553307 on Build GRCh38
rs145180972 Chr.2: 64553270-64553270 on Build GRCh38
rs201431817 Chr.2: 64553381-64553381 on Build GRCh38
rs146619558 Chr.2: 64553399-64553399 on Build GRCh38
rs34934815 Chr.2: 64553332-64553332 on Build GRCh38
rs368338581 Chr.2: 64553379-64553379 on Build GRCh38
rs746081467 Chr.2: 64553281-64553281 on Build GRCh38
rs756442392 Chr.2: 64553285-64553285 on Build GRCh38
rs776192124 Chr.2: 64553325-64553325 on Build GRCh38
rs752089239 Chr.2: 64553362-64553362 on Build GRCh38
rs370571980 Chr.2: 64553309-64553309 on Build GRCh38
rs201878139 Chr.2: 64553370-64553370 on Build GRCh38
rs199744881 Chr.2: 64553334-64553334 on Build GRCh38
rs569518242 Chr.2: 64553337-64553337 on Build GRCh38
rs1009952320 Chr.2: 64553433-64553433 on Build GRCh38
rs770897104 Chr.2: 64553419-64553419 on Build GRCh38
rs763310211 Chr.2: 64553335-64553335 on Build GRCh38
rs963899589 Chr.2: 64553276-64553276 on Build GRCh38
rs765583673 Chr.2: 64553244-64553244 on Build GRCh38
rs140720404 Chr.2: 64553369-64553369 on Build GRCh38
rs758155807 Chr.2: 64553387-64553387 on Build GRCh38
rs770416781 Chr.2: 64553321-64553321 on Build GRCh38
rs368784485 Chr.2: 64553259-64553259 on Build GRCh38
rs945885430 Chr.2: 64553315-64553315 on Build GRCh38
rs752012281 Chr.2: 64553263-64553263 on Build GRCh38
rs757344981 Chr.2: 64553269-64553269 on Build GRCh38
rs773136683 Chr.2: 64553311-64553311 on Build GRCh38

View Full Product Details