Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74722073-74722346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1A1
Entrez Gene ID
1543
Gene Name
cytochrome P450 family 1 subfamily A member 1
Gene Aliases
AHH, AHRR, CP11, CYP1, CYPIA1, P1-450, P450-C, P450DX
UniGene
Hs.72912
Gene Chromosome Location
Chr.15: 74719542-74725610 on Build GRCh38
Transcripts
NM_000499.4 NM_001319216.1 NM_001319217.1 XM_017021953.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7363498 c.793A>G p.M265V Chr.15: 74722305-74722305 on Build GRCh38
7332220 c.810C>A p.Y270* Chr.15: 74722288-74722288 on Build GRCh38
5822429 c.825+3A>G p.? Chr.15: 74722270-74722270 on Build GRCh38
701193 c.766G>T p.E256* Chr.15: 74722332-74722332 on Build GRCh38
701194 c.795G>A p.M265I Chr.15: 74722303-74722303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150514993 Chr.15: 74722305-74722305 on Build GRCh38
rs753707799 Chr.15: 74722282-74722282 on Build GRCh38
rs957719969 Chr.15: 74722136-74722136 on Build GRCh38
rs757497550 Chr.15: 74722314-74722314 on Build GRCh38
rs4986879 Chr.15: 74722228-74722228 on Build GRCh38
rs771326375 Chr.15: 74722223-74722223 on Build GRCh38
rs765333534 Chr.15: 74722296-74722296 on Build GRCh38
rs781571950 Chr.15: 74722226-74722226 on Build GRCh38
rs373584096 Chr.15: 74722252-74722252 on Build GRCh38
rs566136019 Chr.15: 74722158-74722158 on Build GRCh38
rs758554936 Chr.15: 74722304-74722304 on Build GRCh38
rs370247242 Chr.15: 74722267-74722267 on Build GRCh38
rs547688842 Chr.15: 74722099-74722099 on Build GRCh38
rs761449549 Chr.15: 74722245-74722245 on Build GRCh38
rs770302203 Chr.15: 74722231-74722231 on Build GRCh38
rs920498654 Chr.15: 74722124-74722124 on Build GRCh38
rs752565232 Chr.15: 74722300-74722300 on Build GRCh38
rs759345870 Chr.15: 74722293-74722293 on Build GRCh38
rs760209890 Chr.15: 74722262-74722262 on Build GRCh38
rs914831452 Chr.15: 74722182-74722182 on Build GRCh38
rs78058484 Chr.15: 74722130-74722130 on Build GRCh38
rs772963090 Chr.15: 74722261-74722261 on Build GRCh38
rs45564835 Chr.15: 74722242-74722242 on Build GRCh38
rs1050231399 Chr.15: 74722166-74722166 on Build GRCh38
rs775912950 Chr.15: 74722236-74722236 on Build GRCh38
rs954272416 Chr.15: 74722207-74722207 on Build GRCh38
rs781336696 Chr.15: 74722315-74722315 on Build GRCh38
rs766089505 Chr.15: 74722275-74722275 on Build GRCh38
rs139496485 Chr.15: 74722306-74722306 on Build GRCh38

View Full Product Details