Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47936923-47937150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGA1
Entrez Gene ID
1259
Gene Name
cyclic nucleotide gated channel alpha 1
Gene Aliases
CNCG, CNCG1, CNG-1, CNG1, RCNC1, RCNCa, RCNCalpha, RP49
UniGene
Hs.1323
Gene Chromosome Location
Chr.4: 47935015-48016718 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Gene Symbol
LOC101927157
Entrez Gene ID
101927157
Gene Name
uncharacterized LOC101927157
Gene Aliases
-
UniGene
Hs.134190
Gene Chromosome Location
Chr.4: 47914227-47990737 on Build GRCh38
Transcripts
NM_000087.3 NM_001142564.1 XM_005248049.4 XM_011513623.2 XM_017007712.1 NR_125879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1328503 c.1381C>A p.L461I Chr.4: 47937113-47937113 on Build GRCh38
6631107 c.1503T>C p.S501S Chr.4: 47936991-47936991 on Build GRCh38
1429894 c.1541G>A p.R514Q Chr.4: 47936953-47936953 on Build GRCh38
310160 c.1366T>C p.Y456H Chr.4: 47937128-47937128 on Build GRCh38
319522 c.1388C>G p.A463G Chr.4: 47937106-47937106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780719604 Chr.4: 47937090-47937090 on Build GRCh38
rs765136329 Chr.4: 47937062-47937062 on Build GRCh38
rs780623075 Chr.4: 47937006-47937006 on Build GRCh38
rs199584830 Chr.4: 47936954-47936954 on Build GRCh38
rs939558963 Chr.4: 47937107-47937107 on Build GRCh38
rs769089133 Chr.4: 47937117-47937117 on Build GRCh38
rs186845150 Chr.4: 47937003-47937003 on Build GRCh38
rs1009901175 Chr.4: 47936956-47936956 on Build GRCh38
rs749821126 Chr.4: 47937113-47937113 on Build GRCh38
rs754395605 Chr.4: 47937004-47937004 on Build GRCh38
rs761368543 Chr.4: 47937026-47937028 on Build GRCh38
rs773232695 Chr.4: 47937052-47937052 on Build GRCh38
rs374880057 Chr.4: 47937019-47937019 on Build GRCh38
rs777356394 Chr.4: 47936986-47936986 on Build GRCh38
rs201553765 Chr.4: 47937075-47937075 on Build GRCh38
rs989549174 Chr.4: 47937023-47937023 on Build GRCh38
rs769177401 Chr.4: 47937029-47937029 on Build GRCh38
rs966680788 Chr.4: 47937054-47937054 on Build GRCh38
rs544588016 Chr.4: 47936957-47936957 on Build GRCh38
rs983435442 Chr.4: 47937089-47937089 on Build GRCh38
rs372770176 Chr.4: 47936983-47936983 on Build GRCh38
rs752477071 Chr.4: 47936971-47936971 on Build GRCh38
rs767503938 Chr.4: 47936953-47936953 on Build GRCh38
rs749947364 Chr.4: 47937091-47937091 on Build GRCh38
rs371736289 Chr.4: 47937017-47937017 on Build GRCh38
rs756925546 Chr.4: 47937078-47937078 on Build GRCh38
rs746381231 Chr.4: 47937011-47937011 on Build GRCh38
rs34250845 Chr.4: 47937100-47937101 on Build GRCh38
rs765077544 Chr.4: 47936958-47936958 on Build GRCh38
rs369806156 Chr.4: 47937036-47937036 on Build GRCh38
rs755667478 Chr.4: 47937097-47937097 on Build GRCh38
rs746668194 Chr.4: 47936994-47936994 on Build GRCh38
rs749616663 Chr.4: 47937021-47937021 on Build GRCh38
rs767624360 Chr.4: 47937051-47937051 on Build GRCh38
rs780381813 Chr.4: 47937108-47937108 on Build GRCh38
rs369602927 Chr.4: 47937066-47937066 on Build GRCh38
rs759487836 Chr.4: 47937061-47937061 on Build GRCh38
rs774652982 Chr.4: 47937035-47937035 on Build GRCh38

View Full Product Details