Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85280317-85280544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX1
Entrez Gene ID
8092
Gene Name
ALX homeobox 1
Gene Aliases
CART1, FND3, HEL23
UniGene
Hs.41683
Gene Chromosome Location
Chr.12: 85280258-85301784 on Build GRCh38
Transcripts
NM_006982.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3955266 c.165C>A p.P55P Chr.12: 85280426-85280426 on Build GRCh38
6598951 c.195G>A p.L65L Chr.12: 85280456-85280456 on Build GRCh38
6587594 c.184C>A p.H62N Chr.12: 85280445-85280445 on Build GRCh38
3813142 c.106G>C p.D36H Chr.12: 85280367-85280367 on Build GRCh38
5551239 c.147C>G p.C49W Chr.12: 85280408-85280408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048636970 Chr.12: 85280448-85280448 on Build GRCh38
rs760284048 Chr.12: 85280372-85280372 on Build GRCh38
rs776648954 Chr.12: 85280490-85280491 on Build GRCh38
rs746804639 Chr.12: 85280495-85280495 on Build GRCh38
rs766955975 Chr.12: 85280409-85280409 on Build GRCh38
rs772972571 Chr.12: 85280447-85280447 on Build GRCh38
rs781280416 Chr.12: 85280347-85280347 on Build GRCh38
rs750883922 Chr.12: 85280399-85280399 on Build GRCh38
rs776678208 Chr.12: 85280502-85280502 on Build GRCh38
rs757462532 Chr.12: 85280494-85280494 on Build GRCh38
rs115596276 Chr.12: 85280452-85280452 on Build GRCh38
rs769939946 Chr.12: 85280353-85280353 on Build GRCh38
rs372544988 Chr.12: 85280370-85280370 on Build GRCh38
rs141968055 Chr.12: 85280408-85280408 on Build GRCh38
rs748403633 Chr.12: 85280352-85280352 on Build GRCh38
rs774149369 Chr.12: 85280362-85280362 on Build GRCh38
rs561233322 Chr.12: 85280461-85280461 on Build GRCh38
rs375884213 Chr.12: 85280524-85280524 on Build GRCh38
rs776213513 Chr.12: 85280385-85280385 on Build GRCh38
rs368289293 Chr.12: 85280427-85280427 on Build GRCh38
rs575941794 Chr.12: 85280515-85280515 on Build GRCh38
rs369826150 Chr.12: 85280375-85280375 on Build GRCh38
rs756389093 Chr.12: 85280422-85280422 on Build GRCh38
rs201314651 Chr.12: 85280341-85280341 on Build GRCh38
rs775308419 Chr.12: 85280432-85280432 on Build GRCh38
rs748172443 Chr.12: 85280413-85280413 on Build GRCh38
rs762520703 Chr.12: 85280512-85280512 on Build GRCh38
rs777754230 Chr.12: 85280472-85280472 on Build GRCh38
rs986158806 Chr.12: 85280395-85280395 on Build GRCh38
rs914521573 Chr.12: 85280407-85280407 on Build GRCh38
rs759945013 Chr.12: 85280516-85280516 on Build GRCh38
rs145944049 Chr.12: 85280451-85280451 on Build GRCh38
rs778019022 Chr.12: 85280486-85280486 on Build GRCh38
rs376329142 Chr.12: 85280373-85280373 on Build GRCh38
rs768579174 Chr.12: 85280434-85280434 on Build GRCh38
rs148136812 Chr.12: 85280386-85280386 on Build GRCh38
rs373333417 Chr.12: 85280518-85280518 on Build GRCh38
rs776338410 Chr.12: 85280438-85280438 on Build GRCh38
rs115440539 Chr.12: 85280443-85280443 on Build GRCh38
rs761403611 Chr.12: 85280441-85280441 on Build GRCh38
rs768003680 Chr.12: 85280465-85280465 on Build GRCh38
rs375614518 Chr.12: 85280426-85280426 on Build GRCh38
rs764969720 Chr.12: 85280398-85280398 on Build GRCh38
rs770562894 Chr.12: 85280514-85280514 on Build GRCh38
rs368906149 Chr.12: 85280361-85280361 on Build GRCh38
rs749536472 Chr.12: 85280487-85280487 on Build GRCh38
rs755474505 Chr.12: 85280411-85280411 on Build GRCh38
rs775260906 Chr.12: 85280371-85280371 on Build GRCh38
rs751887592 Chr.12: 85280457-85280457 on Build GRCh38
rs138946669 Chr.12: 85280433-85280433 on Build GRCh38
rs752844254 Chr.12: 85280471-85280471 on Build GRCh38
rs150692244 Chr.12: 85280410-85280410 on Build GRCh38
rs779440782 Chr.12: 85280509-85280509 on Build GRCh38
rs772806072 Chr.12: 85280510-85280510 on Build GRCh38
rs781138367 Chr.12: 85280412-85280412 on Build GRCh38

View Full Product Details