Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38298405-38298646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL2
Entrez Gene ID
64225
Gene Name
atlastin GTPase 2
Gene Aliases
ARL3IP2, ARL6IP2, atlastin2
UniGene
Hs.594950
Gene Chromosome Location
Chr.2: 38293957-38378579 on Build GRCh38
Transcripts
NM_001135673.2 NM_001308076.1 NM_022374.3 NR_024191.1 XM_011533017.1 XM_011533018.2 XM_011533019.2 XM_011533020.1 XM_011533022.2 XM_011533023.2 XM_017004686.1 XM_017004687.1 XM_017004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3910318 c.1327C>T p.R443C Chr.2: 38298449-38298449 on Build GRCh38
3910317 c.1327C>T p.R443C Chr.2: 38298449-38298449 on Build GRCh38
7319783 c.1220C>T p.A407V Chr.2: 38298502-38298502 on Build GRCh38
1408018 c.1288C>T p.R430C Chr.2: 38298488-38298488 on Build GRCh38
1183930 c.1249A>C p.K417Q Chr.2: 38298527-38298527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561992226 Chr.2: 38298585-38298585 on Build GRCh38
rs748402865 Chr.2: 38298519-38298519 on Build GRCh38
rs753237588 Chr.2: 38298467-38298467 on Build GRCh38
rs771115333 Chr.2: 38298548-38298548 on Build GRCh38
rs749371764 Chr.2: 38298547-38298547 on Build GRCh38
rs7582826 Chr.2: 38298518-38298518 on Build GRCh38
rs759544075 Chr.2: 38298553-38298553 on Build GRCh38
rs147584252 Chr.2: 38298446-38298446 on Build GRCh38
rs373246244 Chr.2: 38298593-38298593 on Build GRCh38
rs752967814 Chr.2: 38298603-38298603 on Build GRCh38
rs959252559 Chr.2: 38298595-38298595 on Build GRCh38
rs564244279 Chr.2: 38298431-38298431 on Build GRCh38
rs984426120 Chr.2: 38298501-38298501 on Build GRCh38
rs529538215 Chr.2: 38298600-38298600 on Build GRCh38
rs748303608 Chr.2: 38298435-38298435 on Build GRCh38
rs752038537 Chr.2: 38298492-38298492 on Build GRCh38
rs769978665 Chr.2: 38298436-38298436 on Build GRCh38
rs142044948 Chr.2: 38298448-38298448 on Build GRCh38
rs753497754 Chr.2: 38298580-38298580 on Build GRCh38
rs116493165 Chr.2: 38298449-38298449 on Build GRCh38
rs777944175 Chr.2: 38298533-38298533 on Build GRCh38
rs569395890 Chr.2: 38298529-38298529 on Build GRCh38
rs749516332 Chr.2: 38298612-38298612 on Build GRCh38
rs370576887 Chr.2: 38298608-38298608 on Build GRCh38
rs369505051 Chr.2: 38298456-38298456 on Build GRCh38
rs760398662 Chr.2: 38298561-38298561 on Build GRCh38
rs559121783 Chr.2: 38298455-38298455 on Build GRCh38
rs758975947 Chr.2: 38298481-38298481 on Build GRCh38
rs771998613 Chr.2: 38298557-38298557 on Build GRCh38
rs774491502 Chr.2: 38298551-38298551 on Build GRCh38
rs376543656 Chr.2: 38298440-38298440 on Build GRCh38
rs767408959 Chr.2: 38298451-38298451 on Build GRCh38
rs775089987 Chr.2: 38298558-38298558 on Build GRCh38
rs752466538 Chr.2: 38298452-38298452 on Build GRCh38
rs976543712 Chr.2: 38298458-38298458 on Build GRCh38
rs377142879 Chr.2: 38298594-38298594 on Build GRCh38
rs755391206 Chr.2: 38298502-38298502 on Build GRCh38
rs61756317 Chr.2: 38298465-38298465 on Build GRCh38
rs752124620 Chr.2: 38298598-38298598 on Build GRCh38
rs780577030 Chr.2: 38298488-38298488 on Build GRCh38

View Full Product Details