Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184178980-184179205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2M1
Entrez Gene ID
1173
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
AP50, CLAPM1, mu2
UniGene
Hs.518460
Gene Chromosome Location
Chr.3: 184174846-184184091 on Build GRCh38
Transcripts
NM_001025205.1 NM_001311198.1 NM_004068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601629 c.231G>T p.M77I Chr.3: 184179013-184179013 on Build GRCh38
5595678 c.243C>T p.F81F Chr.3: 184179025-184179025 on Build GRCh38
1642192 c.220A>G p.N74D Chr.3: 184179002-184179002 on Build GRCh38
6468710 c.331C>T p.L111L Chr.3: 184179113-184179113 on Build GRCh38
1670739 c.295G>T p.E99* Chr.3: 184179077-184179077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772071290 Chr.3: 184179001-184179001 on Build GRCh38
rs773307248 Chr.3: 184179172-184179172 on Build GRCh38
rs796948561 Chr.3: 184179179-184179179 on Build GRCh38
rs527670858 Chr.3: 184179137-184179137 on Build GRCh38
rs763162356 Chr.3: 184179073-184179073 on Build GRCh38
rs11539599 Chr.3: 184179018-184179018 on Build GRCh38
rs375760613 Chr.3: 184179133-184179133 on Build GRCh38
rs1025759162 Chr.3: 184179159-184179159 on Build GRCh38
rs374693436 Chr.3: 184179169-184179169 on Build GRCh38
rs757002374 Chr.3: 184179150-184179150 on Build GRCh38
rs761946127 Chr.3: 184179005-184179005 on Build GRCh38
rs780025816 Chr.3: 184179160-184179160 on Build GRCh38
rs111539857 Chr.3: 184179123-184179123 on Build GRCh38
rs540567642 Chr.3: 184179149-184179149 on Build GRCh38
rs1008270810 Chr.3: 184179180-184179180 on Build GRCh38
rs200520955 Chr.3: 184179153-184179153 on Build GRCh38
rs775327824 Chr.3: 184179004-184179004 on Build GRCh38
rs368164763 Chr.3: 184179143-184179143 on Build GRCh38
rs199554477 Chr.3: 184179162-184179162 on Build GRCh38
rs995446453 Chr.3: 184179044-184179044 on Build GRCh38
rs766685109 Chr.3: 184179082-184179082 on Build GRCh38
rs768041479 Chr.3: 184179136-184179136 on Build GRCh38
rs2231217 Chr.3: 184179178-184179178 on Build GRCh38
rs1134758 Chr.3: 184179098-184179098 on Build GRCh38
rs771827784 Chr.3: 184179154-184179154 on Build GRCh38
rs560148625 Chr.3: 184179043-184179043 on Build GRCh38
rs370947653 Chr.3: 184179134-184179134 on Build GRCh38
rs777442995 Chr.3: 184179144-184179144 on Build GRCh38
rs11539598 Chr.3: 184179019-184179019 on Build GRCh38

View Full Product Details