Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 75206509-75206782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf39
Entrez Gene ID
56905
Gene Name
chromosome 15 open reading frame 39
Gene Aliases
-
UniGene
Hs.17936
Gene Chromosome Location
Chr.15: 75201880-75212169 on Build GRCh38
Transcripts
NM_015492.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5971536 c.610T>G p.S204A Chr.15: 75206658-75206658 on Build GRCh38
1198139 c.604G>T p.D202Y Chr.15: 75206652-75206652 on Build GRCh38
5198455 c.526C>T p.L176L Chr.15: 75206574-75206574 on Build GRCh38
1939035 c.577G>T p.G193W Chr.15: 75206625-75206625 on Build GRCh38
3721021 c.468T>C p.D156D Chr.15: 75206516-75206516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762870859 Chr.15: 75206703-75206703 on Build GRCh38
rs1031646750 Chr.15: 75206717-75206717 on Build GRCh38
rs774882729 Chr.15: 75206645-75206645 on Build GRCh38
rs768973111 Chr.15: 75206637-75206637 on Build GRCh38
rs372532124 Chr.15: 75206747-75206747 on Build GRCh38
rs765148849 Chr.15: 75206551-75206551 on Build GRCh38
rs765934103 Chr.15: 75206691-75206691 on Build GRCh38
rs143072938 Chr.15: 75206533-75206533 on Build GRCh38
rs759021263 Chr.15: 75206698-75206698 on Build GRCh38
rs753578475 Chr.15: 75206534-75206534 on Build GRCh38
rs781395773 Chr.15: 75206712-75206712 on Build GRCh38
rs372183969 Chr.15: 75206552-75206552 on Build GRCh38
rs754575222 Chr.15: 75206541-75206541 on Build GRCh38
rs148131261 Chr.15: 75206721-75206721 on Build GRCh38
rs1045996896 Chr.15: 75206667-75206667 on Build GRCh38
rs752346892 Chr.15: 75206701-75206701 on Build GRCh38
rs369281067 Chr.15: 75206699-75206699 on Build GRCh38
rs751596174 Chr.15: 75206706-75206706 on Build GRCh38
rs749483382 Chr.15: 75206733-75206733 on Build GRCh38
rs756158757 Chr.15: 75206718-75206718 on Build GRCh38
rs147093913 Chr.15: 75206750-75206750 on Build GRCh38
rs772462766 Chr.15: 75206762-75206762 on Build GRCh38
rs777874459 Chr.15: 75206571-75206571 on Build GRCh38
rs564716678 Chr.15: 75206633-75206633 on Build GRCh38
rs367772385 Chr.15: 75206675-75206675 on Build GRCh38
rs1048977016 Chr.15: 75206626-75206626 on Build GRCh38
rs757245578 Chr.15: 75206707-75206707 on Build GRCh38
rs866242565 Chr.15: 75206580-75206580 on Build GRCh38
rs746902353 Chr.15: 75206587-75206587 on Build GRCh38
rs749135337 Chr.15: 75206623-75206623 on Build GRCh38
rs151171215 Chr.15: 75206602-75206602 on Build GRCh38
rs769062985 Chr.15: 75206744-75206744 on Build GRCh38
rs776002065 Chr.15: 75206694-75206694 on Build GRCh38
rs374466401 Chr.15: 75206596-75206596 on Build GRCh38
rs746156594 Chr.15: 75206623-75206623 on Build GRCh38
rs143715170 Chr.15: 75206628-75206628 on Build GRCh38
rs758480550 Chr.15: 75206722-75206722 on Build GRCh38
rs777563887 Chr.15: 75206621-75206624 on Build GRCh38
rs548759069 Chr.15: 75206622-75206622 on Build GRCh38
rs750471501 Chr.15: 75206714-75206714 on Build GRCh38
rs749663017 Chr.15: 75206629-75206629 on Build GRCh38
rs758210261 Chr.15: 75206563-75206563 on Build GRCh38

View Full Product Details