Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 69841189-69841452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK6
Entrez Gene ID
220164
Gene Name
docking protein 6
Gene Aliases
DOK5L, HsT3226
UniGene
Hs.278285
Gene Chromosome Location
Chr.18: 69401027-69849087 on Build GRCh38
Transcripts
NM_152721.5 XM_011525875.2 XM_017025610.1 XM_017025611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6437774 c.887G>C p.R296P Chr.18: 69841274-69841274 on Build GRCh38
1389592 c.923G>A p.S308N Chr.18: 69841310-69841310 on Build GRCh38
6354945 c.941C>T p.P314L Chr.18: 69841328-69841328 on Build GRCh38
4561689 c.894G>A p.Q298Q Chr.18: 69841281-69841281 on Build GRCh38
5388393 c.950G>A p.R317Q Chr.18: 69841337-69841337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780600018 Chr.18: 69841407-69841407 on Build GRCh38
rs757883092 Chr.18: 69841389-69841390 on Build GRCh38
rs759490627 Chr.18: 69841370-69841370 on Build GRCh38
rs200543822 Chr.18: 69841430-69841430 on Build GRCh38
rs1005791556 Chr.18: 69841224-69841224 on Build GRCh38
rs199575857 Chr.18: 69841397-69841397 on Build GRCh38
rs368072664 Chr.18: 69841367-69841367 on Build GRCh38
rs373764392 Chr.18: 69841431-69841431 on Build GRCh38
rs978843273 Chr.18: 69841335-69841335 on Build GRCh38
rs545055642 Chr.18: 69841296-69841296 on Build GRCh38
rs752154009 Chr.18: 69841390-69841390 on Build GRCh38
rs781553155 Chr.18: 69841317-69841317 on Build GRCh38
rs766696627 Chr.18: 69841260-69841260 on Build GRCh38
rs768980813 Chr.18: 69841268-69841268 on Build GRCh38
rs755544608 Chr.18: 69841425-69841425 on Build GRCh38
rs1023459396 Chr.18: 69841333-69841333 on Build GRCh38
rs1016014086 Chr.18: 69841227-69841227 on Build GRCh38
rs780759548 Chr.18: 69841329-69841329 on Build GRCh38
rs764671949 Chr.18: 69841389-69841389 on Build GRCh38
rs757603917 Chr.18: 69841220-69841220 on Build GRCh38
rs767282823 Chr.18: 69841275-69841275 on Build GRCh38
rs764756255 Chr.18: 69841306-69841306 on Build GRCh38
rs762663116 Chr.18: 69841396-69841396 on Build GRCh38
rs750820207 Chr.18: 69841323-69841323 on Build GRCh38
rs770823707 Chr.18: 69841366-69841366 on Build GRCh38
rs748333355 Chr.18: 69841341-69841341 on Build GRCh38
rs750782550 Chr.18: 69841401-69841401 on Build GRCh38
rs756478289 Chr.18: 69841405-69841405 on Build GRCh38
rs139992081 Chr.18: 69841297-69841297 on Build GRCh38
rs199613374 Chr.18: 69841308-69841308 on Build GRCh38
rs758002118 Chr.18: 69841311-69841311 on Build GRCh38
rs1001893393 Chr.18: 69841270-69841270 on Build GRCh38
rs201544676 Chr.18: 69841361-69841361 on Build GRCh38
rs866303972 Chr.18: 69841352-69841352 on Build GRCh38
rs749350031 Chr.18: 69841330-69841330 on Build GRCh38
rs756635026 Chr.18: 69841328-69841328 on Build GRCh38
rs558527260 Chr.18: 69841274-69841274 on Build GRCh38
rs76231229 Chr.18: 69841259-69841259 on Build GRCh38
rs1038207429 Chr.18: 69841234-69841234 on Build GRCh38
rs578008117 Chr.18: 69841276-69841276 on Build GRCh38
rs746328436 Chr.18: 69841236-69841236 on Build GRCh38
rs373935197 Chr.18: 69841273-69841273 on Build GRCh38
rs770208685 Chr.18: 69841252-69841252 on Build GRCh38
rs200475816 Chr.18: 69841408-69841408 on Build GRCh38
rs779144654 Chr.18: 69841336-69841336 on Build GRCh38
rs542676274 Chr.18: 69841327-69841327 on Build GRCh38
rs772244942 Chr.18: 69841346-69841346 on Build GRCh38
rs778977832 Chr.18: 69841319-69841320 on Build GRCh38
rs760654063 Chr.18: 69841283-69841283 on Build GRCh38
rs781433587 Chr.18: 69841232-69841232 on Build GRCh38
rs143357220 Chr.18: 69841334-69841334 on Build GRCh38
rs773042950 Chr.18: 69841349-69841349 on Build GRCh38

View Full Product Details