Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 23703809-23704070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT9
Entrez Gene ID
23597
Gene Name
acyl-CoA thioesterase 9
Gene Aliases
ACATE2, CGI-16, MT-ACT48, MTACT48
UniGene
Hs.298885
Gene Chromosome Location
Chr.X: 23701055-23743290 on Build GRCh38
Transcripts
NM_001033583.2 NM_001037171.1 XM_005274472.2

SNP Detail

SNP ID Chromosome Location
rs774584941 Chr.X: 23703925-23703925 on Build GRCh38
rs201105808 Chr.X: 23703937-23703937 on Build GRCh38
rs781331000 Chr.X: 23703862-23703862 on Build GRCh38
rs755193322 Chr.X: 23703856-23703856 on Build GRCh38
rs750227406 Chr.X: 23704005-23704005 on Build GRCh38
rs1003218285 Chr.X: 23704026-23704026 on Build GRCh38
rs758244410 Chr.X: 23704024-23704024 on Build GRCh38
rs994982709 Chr.X: 23703848-23703848 on Build GRCh38
rs367791680 Chr.X: 23703909-23703909 on Build GRCh38
rs761691139 Chr.X: 23703979-23703979 on Build GRCh38
rs765152797 Chr.X: 23703998-23703998 on Build GRCh38
rs748354251 Chr.X: 23703880-23703880 on Build GRCh38
rs750182350 Chr.X: 23703852-23703852 on Build GRCh38
rs374622718 Chr.X: 23703954-23703954 on Build GRCh38
rs749809021 Chr.X: 23703899-23703899 on Build GRCh38
rs777876069 Chr.X: 23703886-23703886 on Build GRCh38
rs774409265 Chr.X: 23703947-23703947 on Build GRCh38
rs769926537 Chr.X: 23703884-23703884 on Build GRCh38
rs13497 Chr.X: 23703955-23703955 on Build GRCh38
rs778599831 Chr.X: 23703847-23703847 on Build GRCh38
rs776162907 Chr.X: 23703956-23703956 on Build GRCh38
rs200042097 Chr.X: 23703928-23703928 on Build GRCh38

View Full Product Details