Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76859470-76859717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP2
Entrez Gene ID
1466
Gene Name
cysteine and glycine rich protein 2
Gene Aliases
CRP2, LMO5, SmLIM
UniGene
Hs.530904
Gene Chromosome Location
Chr.12: 76858715-76879040 on Build GRCh38
Transcripts
NM_001300965.1 NM_001321.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272939 c.412-7C>A p.? Chr.12: 76859647-76859647 on Build GRCh38
4921325 c.498T>C p.Y166Y Chr.12: 76859554-76859554 on Build GRCh38
4044896 c.434G>A p.R145Q Chr.12: 76859618-76859618 on Build GRCh38
6636959 c.502A>G p.K168E Chr.12: 76859550-76859550 on Build GRCh38
1364291 c.497A>G p.Y166C Chr.12: 76859555-76859555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748973108 Chr.12: 76859655-76859656 on Build GRCh38
rs761744208 Chr.12: 76859524-76859524 on Build GRCh38
rs370453240 Chr.12: 76859505-76859505 on Build GRCh38
rs777112798 Chr.12: 76859664-76859664 on Build GRCh38
rs988063259 Chr.12: 76859590-76859590 on Build GRCh38
rs777784627 Chr.12: 76859566-76859566 on Build GRCh38
rs762454221 Chr.12: 76859676-76859676 on Build GRCh38
rs756006101 Chr.12: 76859562-76859562 on Build GRCh38
rs752493184 Chr.12: 76859559-76859559 on Build GRCh38
rs765420263 Chr.12: 76859525-76859525 on Build GRCh38
rs765203671 Chr.12: 76859684-76859684 on Build GRCh38
rs749249223 Chr.12: 76859579-76859579 on Build GRCh38
rs758688366 Chr.12: 76859536-76859536 on Build GRCh38
rs764859939 Chr.12: 76859616-76859616 on Build GRCh38
rs142363855 Chr.12: 76859606-76859606 on Build GRCh38
rs202057442 Chr.12: 76859554-76859554 on Build GRCh38
rs750610353 Chr.12: 76859531-76859531 on Build GRCh38
rs996630340 Chr.12: 76859510-76859513 on Build GRCh38
rs367865997 Chr.12: 76859651-76859651 on Build GRCh38
rs371789866 Chr.12: 76859688-76859688 on Build GRCh38
rs530241664 Chr.12: 76859589-76859589 on Build GRCh38
rs770651472 Chr.12: 76859663-76859676 on Build GRCh38
rs372307746 Chr.12: 76859526-76859526 on Build GRCh38
rs561967313 Chr.12: 76859619-76859619 on Build GRCh38
rs769024547 Chr.12: 76859656-76859656 on Build GRCh38
rs781370160 Chr.12: 76859565-76859567 on Build GRCh38
rs376687994 Chr.12: 76859647-76859647 on Build GRCh38
rs368109690 Chr.12: 76859503-76859503 on Build GRCh38
rs144841512 Chr.12: 76859602-76859602 on Build GRCh38

View Full Product Details