Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 129526167-129526404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
UniGene
Hs.23751
Gene Chromosome Location
Chr.5: 129460281-129738683 on Build GRCh38
Transcripts
XM_011543246.2 XM_011543247.2 XM_011543248.2 XM_017009174.1 XM_017009176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060437 c.917G>T p.R306I Chr.5: 129526305-129526305 on Build GRCh38
139593 c.938G>A p.R313K Chr.5: 129526326-129526326 on Build GRCh38
1060441 c.970G>T p.E324* Chr.5: 129526358-129526358 on Build GRCh38
3136508 c.955T>A p.Y319N Chr.5: 129526343-129526343 on Build GRCh38
5908467 c.965C>T p.P322L Chr.5: 129526353-129526353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770807319 Chr.5: 129526276-129526276 on Build GRCh38
rs749760197 Chr.5: 129526302-129526302 on Build GRCh38
rs762665058 Chr.5: 129526311-129526311 on Build GRCh38
rs541019239 Chr.5: 129526266-129526266 on Build GRCh38
rs1055479045 Chr.5: 129526218-129526218 on Build GRCh38
rs932768432 Chr.5: 129526326-129526326 on Build GRCh38
rs779035830 Chr.5: 129526380-129526380 on Build GRCh38
rs536698474 Chr.5: 129526205-129526205 on Build GRCh38
rs775065663 Chr.5: 129526308-129526308 on Build GRCh38
rs758981800 Chr.5: 129526286-129526286 on Build GRCh38
rs778933116 Chr.5: 129526235-129526235 on Build GRCh38
rs371024502 Chr.5: 129526289-129526289 on Build GRCh38
rs747987906 Chr.5: 129526241-129526241 on Build GRCh38
rs758023578 Chr.5: 129526368-129526368 on Build GRCh38
rs767816790 Chr.5: 129526317-129526317 on Build GRCh38
rs771819389 Chr.5: 129526244-129526244 on Build GRCh38
rs777620323 Chr.5: 129526256-129526256 on Build GRCh38
rs750780915 Chr.5: 129526321-129526321 on Build GRCh38
rs938813124 Chr.5: 129526202-129526202 on Build GRCh38
rs753968487 Chr.5: 129526357-129526357 on Build GRCh38
rs776150268 Chr.5: 129526277-129526277 on Build GRCh38
rs761282830 Chr.5: 129526329-129526329 on Build GRCh38
rs1011237014 Chr.5: 129526309-129526309 on Build GRCh38
rs1051256692 Chr.5: 129526352-129526352 on Build GRCh38
rs766949011 Chr.5: 129526335-129526335 on Build GRCh38
rs181359769 Chr.5: 129526258-129526258 on Build GRCh38

View Full Product Details