Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60683891-60684122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4519427 c.100G>A p.E34K Chr.17: 60683998-60683998 on Build GRCh38
4519428 c.100G>A p.E34K Chr.17: 60683998-60683998 on Build GRCh38
2793556 c.129T>C p.V43V Chr.17: 60684027-60684027 on Build GRCh38
1246015 c.88_91delCAGT p.Q30fs*7 Chr.17: 60683986-60683989 on Build GRCh38
4269595 c.129T>C p.V43V Chr.17: 60684027-60684027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978425627 Chr.17: 60684007-60684007 on Build GRCh38
rs999768610 Chr.17: 60684049-60684049 on Build GRCh38
rs752180398 Chr.17: 60683959-60683959 on Build GRCh38
rs1030945661 Chr.17: 60684021-60684021 on Build GRCh38
rs770160173 Chr.17: 60683940-60683940 on Build GRCh38
rs756320993 Chr.17: 60684002-60684002 on Build GRCh38
rs770037197 Chr.17: 60683956-60683956 on Build GRCh38
rs965381494 Chr.17: 60683991-60683991 on Build GRCh38
rs374918966 Chr.17: 60683930-60683930 on Build GRCh38
rs759610827 Chr.17: 60684083-60684084 on Build GRCh38
rs752914908 Chr.17: 60683992-60683992 on Build GRCh38
rs773719014 Chr.17: 60683945-60683945 on Build GRCh38
rs761398614 Chr.17: 60683934-60683934 on Build GRCh38
rs767543165 Chr.17: 60683987-60683987 on Build GRCh38
rs759630539 Chr.17: 60683966-60683966 on Build GRCh38
rs373805993 Chr.17: 60683929-60683929 on Build GRCh38
rs372560258 Chr.17: 60684030-60684030 on Build GRCh38
rs780602504 Chr.17: 60684067-60684067 on Build GRCh38
rs376708535 Chr.17: 60683938-60683938 on Build GRCh38
rs774813504 Chr.17: 60684037-60684040 on Build GRCh38
rs746955029 Chr.17: 60684058-60684058 on Build GRCh38
rs1030714932 Chr.17: 60684059-60684059 on Build GRCh38
rs180742204 Chr.17: 60683925-60683925 on Build GRCh38
rs758244318 Chr.17: 60684042-60684042 on Build GRCh38
rs567226913 Chr.17: 60684033-60684033 on Build GRCh38
rs768186894 Chr.17: 60684060-60684060 on Build GRCh38
rs1041269396 Chr.17: 60683995-60683995 on Build GRCh38
rs763499767 Chr.17: 60683952-60683952 on Build GRCh38
rs779959458 Chr.17: 60684047-60684047 on Build GRCh38
rs747774712 Chr.17: 60684086-60684086 on Build GRCh38
rs776312364 Chr.17: 60683931-60683931 on Build GRCh38
rs766981692 Chr.17: 60683953-60683953 on Build GRCh38

View Full Product Details